Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR2F1-AS1 SIRNA Assays ›
          Gene Symbol:
          NR2F1-AS1
          Gene Name:
          NR2F1 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 93409356 - 93581320 on Build GRCh38
          UniGene ID:
          Hs.457407
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n269413

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NR2F1-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_021490.2 4 470
          NR_021491.2 4 470
          NR_109818.1 5 583
          NR_109819.1 4 1145
          NR_109820.1 4 470
          NR_109821.1 4 470
          NR_109823.1 4 467
          NR_109824.1 4 470
          NR_109825.1 5 1250
          GenBank mRNA AK095719.1 - 5 561
          AK124699.1 - 4 1195
          AK311143.1 - 4 369
          AK311323.1 - 4 858
          BC010610.1 - 4 438
          BC018089.1 - 4 447
          BC042879.1 - 4 369
          BC044619.1 - 5 726
          BC048999.1 - 5 726
          BC144330.1 - 4 1745
          BC144331.1 - 4 1745
          BX647510.1 - 3 238
          DA718791.1 - 4 453
          DB257894.1 - 4 369
          HY108556.1 - 4 400
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB503335.1 - 4 366 Chr. 5 - 93544834 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA531679.1 - 4 521 Chr. 5 - 93543968 - 93581370 [-] on Build GRCh38
          GenBankmRNA BM546601.1 - 535 Chr. - - [-] on Build
          GenBankmRNA DA271332.1 - 4 384 Chr. 5 - 93438714 - 93581234 [-] on Build GRCh38
          GenBankmRNA HY136165.1 - 5 427 Chr. 5 - 93553771 - 93581276 [-] on Build GRCh38
          GenBankmRNA DB486010.2 - 4 366 Chr. 5 - 93411386 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA533837.1 - 4 443 Chr. 5 - 93544815 - 93580618 [-] on Build GRCh38
          GenBankmRNA DA363045.1 - 4 514 Chr. 5 - 93553758 - 93580689 [-] on Build GRCh38
          GenBankmRNA DA134670.1 - 4 369 Chr. 5 - 93411404 - 93581219 [-] on Build GRCh38
          GenBankmRNA DA527197.1 - 4 453 Chr. 5 - 93411404 - 93581303 [-] on Build GRCh38
          GenBankmRNA DA706940.1 - 4 452 Chr. 5 - 93544828 - 93581300 [-] on Build GRCh38
          GenBankmRNA DA280798.1 - 4 351 Chr. 5 - 93411352 - 93581199 [-] on Build GRCh38
          GenBankmRNA BX377086.2 - 4 373 Chr. 5 - 93543540 - 93581223 [-] on Build GRCh38
          GenBankmRNA BG701915.1 - 4 613 Chr. 5 - 93501042 - 93585650 [-] on Build GRCh38
          GenBankmRNA DA844094.1 - 4 449 Chr. 5 - 93544836 - 93581297 [-] on Build GRCh38
          GenBankmRNA DA066475.1 - 2 134 Chr. 5 - 93411149 - 93554938 [-] on Build GRCh38
          GenBankmRNA BI669922.1 - 4 374 Chr. 5 - 93411232 - 93581219 [-] on Build GRCh38
          GenBankmRNA DA545967.1 - 4 369 Chr. 5 - 93544816 - 93581219 [-] on Build GRCh38
          GenBankmRNA DB272535.1 - 4 471 Chr. 5 - 93544853 - 93580646 [-] on Build GRCh38
          GenBankmRNA DA518837.1 - 5 561 Chr. 5 - 93553785 - 93581297 [-] on Build GRCh38
          GenBankmRNA BG715542.1 - 4 454 Chr. 5 - 93411271 - 93581297 [-] on Build GRCh38
          GenBankmRNA DA304607.1 - 4 356 Chr. 5 - 93411321 - 93581205 [-] on Build GRCh38
          GenBankmRNA DB498974.1 - 4 366 Chr. 5 - 93544802 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA839566.1 - 3 260 Chr. 5 - 93432362 - 93563432 [-] on Build GRCh38
          GenBankmRNA HY141502.1 - 4 350 Chr. 5 - 93544815 - 93581199 [-] on Build GRCh38
          GenBankmRNA BG707159.1 - 4 374 Chr. 5 - 93411365 - 93581219 [-] on Build GRCh38
          GenBankmRNA DA388648.1 - 4 520 Chr. 5 - 93544889 - 93580695 [-] on Build GRCh38
          GenBankmRNA DA328801.1 - 4 371 Chr. 5 - 93543992 - 93581221 [-] on Build GRCh38
          GenBankmRNA BI820847.1 - 4 420 Chr. 5 - 93342240 - 93580595 [-] on Build GRCh38
          GenBankmRNA DA517345.1 - 4 446 Chr. 5 - 93411434 - 93581295 [-] on Build GRCh38
          GenBankmRNA DB492381.1 - 4 366 Chr. 5 - 93544849 - 93581216 [-] on Build GRCh38
          GenBankmRNA DB475119.2 - 4 444 Chr. 5 - 93411324 - 93581297 [-] on Build GRCh38
          GenBankmRNA DB177914.1 - 4 450 Chr. 5 - 93438700 - 93581300 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0