Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR2F1-AS1 SIRNA Assays ›
          Gene Symbol:
          NR2F1-AS1
          Gene Name:
          NR2F1 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 93409356 - 93581320 on Build GRCh38
          UniGene ID:
          Hs.457407
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n269414

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NR2F1-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_021490.2 2 260
          NR_021491.2 2 260
          NR_109818.1 3 373
          NR_109819.1 2 935
          NR_109820.1 2 260
          NR_109821.1 2 260
          NR_109822.1 2 935
          NR_109823.1 2 257
          NR_109824.1 2 260
          NR_109825.1 3 1040
          GenBank mRNA AK095719.1 - 3 351
          AK124699.1 - 2 985
          AK296281.1 - 2 622
          AK311143.1 - 2 159
          AK311323.1 - 2 648
          BC010610.1 - 2 228
          BC018089.1 - 2 237
          BC042879.1 - 2 159
          BC044619.1 - 3 516
          BC048999.1 - 3 516
          BC144330.1 - 2 1535
          BC144331.1 - 2 1535
          BX647510.1 - 1 28
          DA718791.1 - 2 243
          DB257894.1 - 2 159
          HY108556.1 - 2 190
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB503335.1 - 2 156 Chr. 5 - 93544834 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA531679.1 - 2 311 Chr. 5 - 93543968 - 93581370 [-] on Build GRCh38
          GenBankmRNA BI463026.1 - 3 523 Chr. 5 - 93554872 - 93570826 [-] on Build GRCh38
          GenBankmRNA DA530156.1 - 2 509 Chr. 5 - 93554928 - 93580891 [-] on Build GRCh38
          GenBankmRNA DB045052.1 - 3 450 Chr. 5 - 93554901 - 93570793 [-] on Build GRCh38
          GenBankmRNA BM546601.1 - 325 Chr. - - [-] on Build
          GenBankmRNA DA271332.1 - 2 174 Chr. 5 - 93438714 - 93581234 [-] on Build GRCh38
          GenBankmRNA HY136165.1 - 3 217 Chr. 5 - 93553771 - 93581276 [-] on Build GRCh38
          GenBankmRNA DB173308.1 - 2 410 Chr. 5 - 93553837 - 93580795 [-] on Build GRCh38
          GenBankmRNA DB486010.2 - 2 156 Chr. 5 - 93411386 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA533837.1 - 2 233 Chr. 5 - 93544815 - 93580618 [-] on Build GRCh38
          GenBankmRNA DA363045.1 - 2 304 Chr. 5 - 93553758 - 93580689 [-] on Build GRCh38
          GenBankmRNA DA134670.1 - 2 159 Chr. 5 - 93411404 - 93581219 [-] on Build GRCh38
          GenBankmRNA BF530372.1 - 229 Chr. - - [-] on Build
          GenBankmRNA DA527197.1 - 2 243 Chr. 5 - 93411404 - 93581303 [-] on Build GRCh38
          GenBankmRNA DA706940.1 - 2 242 Chr. 5 - 93544828 - 93581300 [-] on Build GRCh38
          GenBankmRNA DA280798.1 - 2 140 Chr. 5 - 93411352 - 93581199 [-] on Build GRCh38
          GenBankmRNA BX377086.2 - 2 163 Chr. 5 - 93543540 - 93581223 [-] on Build GRCh38
          GenBankmRNA BG701915.1 - 2 404 Chr. 5 - 93501042 - 93585650 [-] on Build GRCh38
          GenBankmRNA DA844094.1 - 2 239 Chr. 5 - 93544836 - 93581297 [-] on Build GRCh38
          GenBankmRNA AA375334.1 - 2 108 Chr. 5 - 93553829 - 93581168 [-] on Build GRCh38
          GenBankmRNA BI669922.1 - 2 164 Chr. 5 - 93411232 - 93581219 [-] on Build GRCh38
          GenBankmRNA DA518837.1 - 3 351 Chr. 5 - 93553785 - 93581297 [-] on Build GRCh38
          GenBankmRNA DA545967.1 - 2 159 Chr. 5 - 93544816 - 93581219 [-] on Build GRCh38
          GenBankmRNA DB272535.1 - 2 261 Chr. 5 - 93544853 - 93580646 [-] on Build GRCh38
          GenBankmRNA DA304607.1 - 2 146 Chr. 5 - 93411321 - 93581205 [-] on Build GRCh38
          GenBankmRNA DB498974.1 - 2 156 Chr. 5 - 93544802 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA839566.1 - 1 50 Chr. 5 - 93432362 - 93563432 [-] on Build GRCh38
          GenBankmRNA HY141502.1 - 2 140 Chr. 5 - 93544815 - 93581199 [-] on Build GRCh38
          GenBankmRNA BG707159.1 - 2 164 Chr. 5 - 93411365 - 93581219 [-] on Build GRCh38
          GenBankmRNA DA388648.1 - 2 310 Chr. 5 - 93544889 - 93580695 [-] on Build GRCh38
          GenBankmRNA DA328801.1 - 2 161 Chr. 5 - 93543992 - 93581221 [-] on Build GRCh38
          GenBankmRNA DN993098.1 - 2 361 Chr. 5 - 93553814 - 93580737 [-] on Build GRCh38
          GenBankmRNA BI820847.1 - 2 210 Chr. 5 - 93342240 - 93580595 [-] on Build GRCh38
          GenBankmRNA DA517345.1 - 2 236 Chr. 5 - 93411434 - 93581295 [-] on Build GRCh38
          GenBankmRNA DB492381.1 - 2 156 Chr. 5 - 93544849 - 93581216 [-] on Build GRCh38
          GenBankmRNA BX444443.2 - 2 799 Chr. 5 - 93553821 - 93581185 [-] on Build GRCh38
          GenBankmRNA DB475119.2 - 2 234 Chr. 5 - 93411324 - 93581297 [-] on Build GRCh38
          GenBankmRNA DB177914.1 - 2 240 Chr. 5 - 93438700 - 93581300 [-] on Build GRCh38
          GenBankmRNA DB030965.1 - 3 419 Chr. 5 - 93554880 - 93570722 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-bnpf6:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0