Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR2F1-AS1 SIRNA Assays ›
          Gene Symbol:
          NR2F1-AS1
          Gene Name:
          NR2F1 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 93409356 - 93581320 on Build GRCh38
          UniGene ID:
          Hs.457407
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n269415

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NR2F1-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_021490.2 3 317
          NR_021491.2 3 317
          NR_109818.1 4 430
          NR_109819.1 3 992
          NR_109820.1 3 317
          NR_109821.1 3 317
          NR_109823.1 3 314
          NR_109824.1 3 317
          NR_109825.1 4 1097
          GenBank mRNA AK095719.1 - 4 408
          AK124699.1 - 3 1042
          AK311143.1 - 3 216
          AK311323.1 - 3 705
          BC010610.1 - 3 285
          BC018089.1 - 3 294
          BC042879.1 - 3 216
          BC044619.1 - 4 573
          BC048999.1 - 4 573
          BC144330.1 - 3 1592
          BC144331.1 - 3 1592
          BX647510.1 - 2 85
          DA718791.1 - 3 300
          DB257894.1 - 3 216
          HY108556.1 - 3 247
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB503335.1 - 3 213 Chr. 5 - 93544834 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA531679.1 - 3 368 Chr. 5 - 93543968 - 93581370 [-] on Build GRCh38
          GenBankmRNA BI463026.1 - 4 580 Chr. 5 - 93554872 - 93570826 [-] on Build GRCh38
          GenBankmRNA DA530156.1 - 3 566 Chr. 5 - 93554928 - 93580891 [-] on Build GRCh38
          GenBankmRNA DB045052.1 - 4 507 Chr. 5 - 93554901 - 93570793 [-] on Build GRCh38
          GenBankmRNA BM546601.1 - 382 Chr. - - [-] on Build
          GenBankmRNA DA271332.1 - 3 231 Chr. 5 - 93438714 - 93581234 [-] on Build GRCh38
          GenBankmRNA HY136165.1 - 4 274 Chr. 5 - 93553771 - 93581276 [-] on Build GRCh38
          GenBankmRNA DB486010.2 - 3 213 Chr. 5 - 93411386 - 93581216 [-] on Build GRCh38
          GenBankmRNA DB173308.1 - 3 467 Chr. 5 - 93553837 - 93580795 [-] on Build GRCh38
          GenBankmRNA DA533837.1 - 3 290 Chr. 5 - 93544815 - 93580618 [-] on Build GRCh38
          GenBankmRNA DA363045.1 - 3 361 Chr. 5 - 93553758 - 93580689 [-] on Build GRCh38
          GenBankmRNA DA134670.1 - 3 216 Chr. 5 - 93411404 - 93581219 [-] on Build GRCh38
          GenBankmRNA BF530372.1 - 289 Chr. - - [-] on Build
          GenBankmRNA DA527197.1 - 3 300 Chr. 5 - 93411404 - 93581303 [-] on Build GRCh38
          GenBankmRNA DA706940.1 - 3 299 Chr. 5 - 93544828 - 93581300 [-] on Build GRCh38
          GenBankmRNA DA280798.1 - 3 198 Chr. 5 - 93411352 - 93581199 [-] on Build GRCh38
          GenBankmRNA BX377086.2 - 3 220 Chr. 5 - 93543540 - 93581223 [-] on Build GRCh38
          GenBankmRNA BG701915.1 - 3 461 Chr. 5 - 93501042 - 93585650 [-] on Build GRCh38
          GenBankmRNA DA844094.1 - 3 296 Chr. 5 - 93544836 - 93581297 [-] on Build GRCh38
          GenBankmRNA AA375334.1 - 3 165 Chr. 5 - 93553829 - 93581168 [-] on Build GRCh38
          GenBankmRNA BI669922.1 - 3 221 Chr. 5 - 93411232 - 93581219 [-] on Build GRCh38
          GenBankmRNA DB272535.1 - 3 318 Chr. 5 - 93544853 - 93580646 [-] on Build GRCh38
          GenBankmRNA DA518837.1 - 4 408 Chr. 5 - 93553785 - 93581297 [-] on Build GRCh38
          GenBankmRNA DA545967.1 - 3 216 Chr. 5 - 93544816 - 93581219 [-] on Build GRCh38
          GenBankmRNA BG715542.1 - 3 301 Chr. 5 - 93411271 - 93581297 [-] on Build GRCh38
          GenBankmRNA DA304607.1 - 3 203 Chr. 5 - 93411321 - 93581205 [-] on Build GRCh38
          GenBankmRNA DB498974.1 - 3 213 Chr. 5 - 93544802 - 93581216 [-] on Build GRCh38
          GenBankmRNA DA839566.1 - 2 107 Chr. 5 - 93432362 - 93563432 [-] on Build GRCh38
          GenBankmRNA HY141502.1 - 3 197 Chr. 5 - 93544815 - 93581199 [-] on Build GRCh38
          GenBankmRNA BG707159.1 - 3 221 Chr. 5 - 93411365 - 93581219 [-] on Build GRCh38
          GenBankmRNA DA328801.1 - 3 218 Chr. 5 - 93543992 - 93581221 [-] on Build GRCh38
          GenBankmRNA DA388648.1 - 3 367 Chr. 5 - 93544889 - 93580695 [-] on Build GRCh38
          GenBankmRNA DN993098.1 - 3 418 Chr. 5 - 93553814 - 93580737 [-] on Build GRCh38
          GenBankmRNA BI820847.1 - 3 267 Chr. 5 - 93342240 - 93580595 [-] on Build GRCh38
          GenBankmRNA DA517345.1 - 3 293 Chr. 5 - 93411434 - 93581295 [-] on Build GRCh38
          GenBankmRNA DB492381.1 - 3 213 Chr. 5 - 93544849 - 93581216 [-] on Build GRCh38
          GenBankmRNA DB475119.2 - 3 291 Chr. 5 - 93411324 - 93581297 [-] on Build GRCh38
          GenBankmRNA BX444443.2 - 3 856 Chr. 5 - 93553821 - 93581185 [-] on Build GRCh38
          GenBankmRNA DB177914.1 - 3 297 Chr. 5 - 93438700 - 93581300 [-] on Build GRCh38
          GenBankmRNA DB030965.1 - 4 476 Chr. 5 - 93554880 - 93570722 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0