Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG15 SIRNA Assays ›
          Gene Symbol:
          SNHG15
          Gene Name:
          small nucleolar RNA host gene 15
          Gene Aliases:
          C7orf40, Linc-Myo1g, MYO1GUT
          Chromosome Location:
          Chr.7: 44983028 - 44986660 on Build GRCh38
          UniGene ID:
          Hs.25892
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270567

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003697.1 5 580
          GenBank mRNA AK096179.1 - 2 2888
          BC092459.1 - 5 580
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM772795.1 - 5 544 Chr. 7 - 44983206 - 44986617 [-] on Build GRCh38
          GenBankmRNA BE889371.1 - 5 486 Chr. 7 - 44983041 - 44986566 [-] on Build GRCh38
          GenBankmRNA BM685499.1 - 5 537 Chr. 7 - 44983169 - 44986611 [-] on Build GRCh38
          GenBankmRNA HX788853.1 - 5 569 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA BU537835.1 - 5 588 Chr. 7 - 44983141 - 44986668 [-] on Build GRCh38
          GenBankmRNA AB371433.1 - 5 558 Chr. 7 - 44983029 - 44986637 [-] on Build GRCh38
          GenBankmRNA BY996199.1 - 5 534 Chr. 7 - 44983146 - 44986608 [-] on Build GRCh38
          GenBankmRNA BE888491.1 - 524 Chr. - - [-] on Build
          GenBankmRNA CD641901.1 - 4 496 Chr. 7 - 44983113 - 44986650 [-] on Build GRCh38
          GenBankmRNA BF304363.1 - 506 Chr. - - [-] on Build
          GenBankmRNA BQ059848.1 - 409 Chr. - - [-] on Build
          GenBankmRNA BM720519.1 - 3 349 Chr. 7 - 44983133 - 44984408 [-] on Build GRCh38
          GenBankmRNA BG829322.1 - 5 579 Chr. 7 - 44983089 - 44986658 [-] on Build GRCh38
          GenBankmRNA BF308868.1 - 5 505 Chr. 7 - 44983055 - 44986586 [-] on Build GRCh38
          GenBankmRNA BI907654.1 - 4 506 Chr. 7 - 44983026 - 44986656 [-] on Build GRCh38
          GenBankmRNA BG026375.1 - 366 Chr. - - [-] on Build
          GenBankmRNA CD523041.1 - 2 148 Chr. 7 - 44983019 - 44983896 [-] on Build GRCh38
          GenBankmRNA CN265181.1 - 2 326 Chr. 7 - 44983033 - 44984039 [-] on Build GRCh38
          GenBankmRNA BM764510.1 - 5 544 Chr. 7 - 44983184 - 44986617 [-] on Build GRCh38
          GenBankmRNA AA488149.1 - 3 342 Chr. 7 - 44983085 - 44986570 [-] on Build GRCh38
          GenBankmRNA BE206230.1 - 5 481 Chr. 7 - 44983229 - 44986559 [-] on Build GRCh38
          GenBankmRNA DB560851.1 - 2 225 Chr. 7 - 44983026 - 44983974 [-] on Build GRCh38
          GenBankmRNA BF306650.1 - 510 Chr. - - [-] on Build
          GenBankmRNA CF127244.1 - 224 Chr. - - [-] on Build
          GenBankmRNA AW245923.1 - 5 585 Chr. 7 - 44983229 - 44986640 [-] on Build GRCh38
          GenBankmRNA BF204739.1 - 5 579 Chr. 7 - 44983092 - 44986658 [-] on Build GRCh38
          GenBankmRNA BM825153.1 - 2 355 Chr. 7 - 44983026 - 44984104 [-] on Build GRCh38
          GenBankmRNA BU195547.1 - 5 528 Chr. 7 - 44983030 - 44986605 [-] on Build GRCh38
          GenBankmRNA BG491747.1 - 5 602 Chr. 7 - 44983061 - 44986681 [-] on Build GRCh38
          GenBankmRNA HX785737.1 - 5 569 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA BG748946.1 - 5 545 Chr. 7 - 44983065 - 44986624 [-] on Build GRCh38
          GenBankmRNA BU502253.1 - 5 527 Chr. 7 - 44983028 - 44986604 [-] on Build GRCh38
          GenBankmRNA BQ434731.1 - 5 558 Chr. 7 - 44983024 - 44986638 [-] on Build GRCh38
          GenBankmRNA BG548028.1 - 5 588 Chr. 7 - 44983041 - 44986665 [-] on Build GRCh38
          GenBankmRNA BU535553.1 - 5 642 Chr. 7 - 44983144 - 44986583 [-] on Build GRCh38
          GenBankmRNA CN271520.1 - 4 450 Chr. 7 - 44983122 - 44986600 [-] on Build GRCh38
          GenBankmRNA DB553836.1 - 2 210 Chr. 7 - 44983026 - 44983961 [-] on Build GRCh38
          GenBankmRNA CD639046.1 - 4 496 Chr. 7 - 44983079 - 44986650 [-] on Build GRCh38
          GenBankmRNA BG677227.1 - 5 517 Chr. 7 - 44983131 - 44986597 [-] on Build GRCh38
          GenBankmRNA CD640711.1 - 5 530 Chr. 7 - 44983119 - 44986607 [-] on Build GRCh38
          GenBankmRNA BF314122.1 - 5 505 Chr. 7 - 44983046 - 44986586 [-] on Build GRCh38
          GenBankmRNA BF317113.1 - 5 574 Chr. 7 - 44983055 - 44986653 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0