Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG15 SIRNA Assays ›
          Gene Symbol:
          SNHG15
          Gene Name:
          small nucleolar RNA host gene 15
          Gene Aliases:
          C7orf40, Linc-Myo1g, MYO1GUT
          Chromosome Location:
          Chr.7: 44983028 - 44986660 on Build GRCh38
          UniGene ID:
          Hs.25892
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270570

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003697.1 5 609
          GenBank mRNA AK096179.1 - 2 2917
          BC092459.1 - 5 609
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM772795.1 - 5 573 Chr. 7 - 44983206 - 44986617 [-] on Build GRCh38
          GenBankmRNA BE889371.1 - 5 515 Chr. 7 - 44983041 - 44986566 [-] on Build GRCh38
          GenBankmRNA BM685499.1 - 5 566 Chr. 7 - 44983169 - 44986611 [-] on Build GRCh38
          GenBankmRNA HX788853.1 - 5 598 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA BU537835.1 - 5 617 Chr. 7 - 44983141 - 44986668 [-] on Build GRCh38
          GenBankmRNA AB371433.1 - 5 587 Chr. 7 - 44983029 - 44986637 [-] on Build GRCh38
          GenBankmRNA BY996199.1 - 5 563 Chr. 7 - 44983146 - 44986608 [-] on Build GRCh38
          GenBankmRNA BE888491.1 - 553 Chr. - - [-] on Build
          GenBankmRNA CD641901.1 - 4 525 Chr. 7 - 44983113 - 44986650 [-] on Build GRCh38
          GenBankmRNA BF304363.1 - 535 Chr. - - [-] on Build
          GenBankmRNA BQ059848.1 - 438 Chr. - - [-] on Build
          GenBankmRNA DB563548.1 - 2 251 Chr. 7 - 44983026 - 44983970 [-] on Build GRCh38
          GenBankmRNA BG829322.1 - 5 608 Chr. 7 - 44983089 - 44986658 [-] on Build GRCh38
          GenBankmRNA BM720519.1 - 3 378 Chr. 7 - 44983133 - 44984408 [-] on Build GRCh38
          GenBankmRNA BF308868.1 - 5 534 Chr. 7 - 44983055 - 44986586 [-] on Build GRCh38
          GenBankmRNA BI907654.1 - 4 535 Chr. 7 - 44983026 - 44986656 [-] on Build GRCh38
          GenBankmRNA BG026375.1 - 395 Chr. - - [-] on Build
          GenBankmRNA CD523041.1 - 2 177 Chr. 7 - 44983019 - 44983896 [-] on Build GRCh38
          GenBankmRNA CN265181.1 - 2 355 Chr. 7 - 44983033 - 44984039 [-] on Build GRCh38
          GenBankmRNA BM764510.1 - 5 573 Chr. 7 - 44983184 - 44986617 [-] on Build GRCh38
          GenBankmRNA AA488149.1 - 3 371 Chr. 7 - 44983085 - 44986570 [-] on Build GRCh38
          GenBankmRNA DB560851.1 - 2 254 Chr. 7 - 44983026 - 44983974 [-] on Build GRCh38
          GenBankmRNA BF306650.1 - 539 Chr. - - [-] on Build
          GenBankmRNA CF127244.1 - 253 Chr. - - [-] on Build
          GenBankmRNA BE312499.1 - 5 514 Chr. 7 - 44983106 - 44986566 [-] on Build GRCh38
          GenBankmRNA BF204739.1 - 5 608 Chr. 7 - 44983092 - 44986658 [-] on Build GRCh38
          GenBankmRNA BM825153.1 - 2 384 Chr. 7 - 44983026 - 44984104 [-] on Build GRCh38
          GenBankmRNA BU195547.1 - 5 557 Chr. 7 - 44983030 - 44986605 [-] on Build GRCh38
          GenBankmRNA BG491747.1 - 5 630 Chr. 7 - 44983061 - 44986681 [-] on Build GRCh38
          GenBankmRNA HX785737.1 - 5 598 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA BG748946.1 - 5 574 Chr. 7 - 44983065 - 44986624 [-] on Build GRCh38
          GenBankmRNA BU502253.1 - 5 556 Chr. 7 - 44983028 - 44986604 [-] on Build GRCh38
          GenBankmRNA BQ434731.1 - 5 587 Chr. 7 - 44983024 - 44986638 [-] on Build GRCh38
          GenBankmRNA BU535553.1 - 5 671 Chr. 7 - 44983144 - 44986583 [-] on Build GRCh38
          GenBankmRNA CN271520.1 - 4 479 Chr. 7 - 44983122 - 44986600 [-] on Build GRCh38
          GenBankmRNA DB553836.1 - 2 239 Chr. 7 - 44983026 - 44983961 [-] on Build GRCh38
          GenBankmRNA CD639046.1 - 4 525 Chr. 7 - 44983079 - 44986650 [-] on Build GRCh38
          GenBankmRNA BG677227.1 - 5 546 Chr. 7 - 44983131 - 44986597 [-] on Build GRCh38
          GenBankmRNA CD640711.1 - 5 559 Chr. 7 - 44983119 - 44986607 [-] on Build GRCh38
          GenBankmRNA BF314122.1 - 5 534 Chr. 7 - 44983046 - 44986586 [-] on Build GRCh38
          GenBankmRNA BG927882.1 - 5 651 Chr. 7 - 44983170 - 44986619 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0