Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG15 SIRNA Assays ›
          Gene Symbol:
          SNHG15
          Gene Name:
          small nucleolar RNA host gene 15
          Gene Aliases:
          C7orf40, Linc-Myo1g, MYO1GUT
          Chromosome Location:
          Chr.7: 44983028 - 44986660 on Build GRCh38
          UniGene ID:
          Hs.25892
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270573

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003697.1 3, 4 282
          GenBank mRNA BC092459.1 - 3, 4 282
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AA524713.1 - 3, 4 374 Chr. 7 - 44983904 - 44986757 [-] on Build GRCh38
          GenBankmRNA R70423.1 - 248 Chr. - - [-] on Build
          GenBankmRNA BE889371.1 - 3, 4 188 Chr. 7 - 44983041 - 44986566 [-] on Build GRCh38
          GenBankmRNA W57948.1 - 253 Chr. - - [-] on Build
          GenBankmRNA BM710323.1 - 3, 4 303 Chr. 7 - 44983870 - 44986681 [-] on Build GRCh38
          GenBankmRNA HX788853.1 - 3, 4 271 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA BE888491.1 - 226 Chr. - - [-] on Build
          GenBankmRNA BF304363.1 - 208 Chr. - - [-] on Build
          GenBankmRNA BM720519.1 - 1, 2 51 Chr. 7 - 44983133 - 44984408 [-] on Build GRCh38
          GenBankmRNA BE206230.1 - 3, 4 183 Chr. 7 - 44983229 - 44986559 [-] on Build GRCh38
          GenBankmRNA AA366086.1 - 3, 4 261 Chr. 7 - 44983993 - 44986638 [-] on Build GRCh38
          GenBankmRNA BF131634.1 - 222 Chr. - - [-] on Build
          GenBankmRNA BF306650.1 - 212 Chr. - - [-] on Build
          GenBankmRNA AW245923.1 - 3, 4 287 Chr. 7 - 44983229 - 44986640 [-] on Build GRCh38
          GenBankmRNA BQ325942.1 - 3, 4 310 Chr. 7 - 44983294 - 44986665 [-] on Build GRCh38
          GenBankmRNA BF204739.1 - 3, 4 281 Chr. 7 - 44983092 - 44986658 [-] on Build GRCh38
          GenBankmRNA DB490500.1 - 282 Chr. 7 - 44984357 - 44986660 [-] on Build GRCh38
          GenBankmRNA BU195547.1 - 3, 4 230 Chr. 7 - 44983030 - 44986605 [-] on Build GRCh38
          GenBankmRNA BG491747.1 - 3, 4 304 Chr. 7 - 44983061 - 44986681 [-] on Build GRCh38
          GenBankmRNA CF128961.1 - 3, 4 247 Chr. 7 - 44983832 - 44986625 [-] on Build GRCh38
          GenBankmRNA BU502253.1 - 3, 4 229 Chr. 7 - 44983028 - 44986604 [-] on Build GRCh38
          GenBankmRNA AA191160.1 - 1, 2 50 Chr. 7 - 44983842 - 44984407 [-] on Build GRCh38
          GenBankmRNA BE733369.1 - 3, 4 456 Chr. 7 - 44983106 - 44986834 [-] on Build GRCh38
          GenBankmRNA CN482323.1 - 3, 4 369 Chr. 7 - 44983304 - 44986608 [-] on Build GRCh38
          GenBankmRNA BM744843.1 - 3, 4 246 Chr. 7 - 44983285 - 44986617 [-] on Build GRCh38
          GenBankmRNA BG677227.1 - 3, 4 219 Chr. 7 - 44983131 - 44986597 [-] on Build GRCh38
          GenBankmRNA CD640711.1 - 3, 4 232 Chr. 7 - 44983119 - 44986607 [-] on Build GRCh38
          GenBankmRNA BG829241.1 - 3, 4 281 Chr. 7 - 44983237 - 44986619 [-] on Build GRCh38
          GenBankmRNA BG927882.1 - 3, 4 324 Chr. 7 - 44983170 - 44986619 [-] on Build GRCh38
          GenBankmRNA BM772795.1 - 3, 4 246 Chr. 7 - 44983206 - 44986617 [-] on Build GRCh38
          GenBankmRNA AA252002.1 - 3, 4 258 Chr. 7 - 44983817 - 44986638 [-] on Build GRCh38
          GenBankmRNA BM758831.1 - 3, 4 246 Chr. 7 - 44983894 - 44986624 [-] on Build GRCh38
          GenBankmRNA DB484794.1 - 3, 4 282 Chr. 7 - 44983890 - 44986660 [-] on Build GRCh38
          GenBankmRNA DW440519.1 - 1, 2 81 Chr. 7 - 44983244 - 44984422 [-] on Build GRCh38
          GenBankmRNA BM685499.1 - 3, 4 240 Chr. 7 - 44983169 - 44986611 [-] on Build GRCh38
          GenBankmRNA AB371433.1 - 3, 4 260 Chr. 7 - 44983029 - 44986637 [-] on Build GRCh38
          GenBankmRNA BU537835.1 - 3, 4 290 Chr. 7 - 44983141 - 44986668 [-] on Build GRCh38
          GenBankmRNA BY996199.1 - 3, 4 236 Chr. 7 - 44983146 - 44986608 [-] on Build GRCh38
          GenBankmRNA BU941648.1 - 3, 4 432 Chr. 7 - 44983210 - 44986670 [-] on Build GRCh38
          GenBankmRNA CD641901.1 - 2, 3 198 Chr. 7 - 44983113 - 44986650 [-] on Build GRCh38
          GenBankmRNA AW328480.1 - 3, 4 197 Chr. 7 - 44983242 - 44986566 [-] on Build GRCh38
          GenBankmRNA AW960033.1 - 3, 4 259 Chr. 7 - 44983173 - 44986637 [-] on Build GRCh38
          GenBankmRNA BM887994.1 - 3, 4 210 Chr. 7 - 44983268 - 44986588 [-] on Build GRCh38
          GenBankmRNA DB492530.1 - 3, 4 282 Chr. 7 - 44983886 - 44986660 [-] on Build GRCh38
          GenBankmRNA BG829322.1 - 3, 4 281 Chr. 7 - 44983089 - 44986658 [-] on Build GRCh38
          GenBankmRNA BF308868.1 - 3, 4 207 Chr. 7 - 44983055 - 44986586 [-] on Build GRCh38
          GenBankmRNA BM764510.1 - 3, 4 246 Chr. 7 - 44983184 - 44986617 [-] on Build GRCh38
          GenBankmRNA N35269.1 - 3, 4 241 Chr. 7 - 44983929 - 44986621 [-] on Build GRCh38
          GenBankmRNA BE312499.1 - 3, 4 188 Chr. 7 - 44983106 - 44986566 [-] on Build GRCh38
          GenBankmRNA BM773240.1 - 3, 4 246 Chr. 7 - 44983897 - 44986617 [-] on Build GRCh38
          GenBankmRNA AA258914.1 - 3, 4 230 Chr. 7 - 44983817 - 44986598 [-] on Build GRCh38
          GenBankmRNA HX785737.1 - 3, 4 271 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA BG748946.1 - 3, 4 247 Chr. 7 - 44983065 - 44986624 [-] on Build GRCh38
          GenBankmRNA AA306794.1 - 3, 4 217 Chr. 7 - 44983244 - 44986595 [-] on Build GRCh38
          GenBankmRNA BG548028.1 - 3, 4 288 Chr. 7 - 44983041 - 44986665 [-] on Build GRCh38
          GenBankmRNA BQ434731.1 - 3, 4 260 Chr. 7 - 44983024 - 44986638 [-] on Build GRCh38
          GenBankmRNA CD522132.1 - 285 Chr. - - [-] on Build
          GenBankmRNA BU535553.1 - 3, 4 344 Chr. 7 - 44983144 - 44986583 [-] on Build GRCh38
          GenBankmRNA CD639046.1 - 2, 3 198 Chr. 7 - 44983079 - 44986650 [-] on Build GRCh38
          GenBankmRNA AA281810.1 - 3, 4 259 Chr. 7 - 44983942 - 44986638 [-] on Build GRCh38
          GenBankmRNA BF317113.1 - 3, 4 276 Chr. 7 - 44983055 - 44986653 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xjq9k:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0