Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG15 SIRNA Assays ›
          Gene Symbol:
          SNHG15
          Gene Name:
          small nucleolar RNA host gene 15
          Gene Aliases:
          C7orf40, Linc-Myo1g, MYO1GUT
          Chromosome Location:
          Chr.7: 44983028 - 44986660 on Build GRCh38
          UniGene ID:
          Hs.25892
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n270574

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003697.1 5 522
          GenBank mRNA AK096179.1 - 2 2830
          BC092459.1 - 5 522
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BE889371.1 - 5 428 Chr. 7 - 44983041 - 44986566 [-] on Build GRCh38
          GenBankmRNA HX788853.1 - 5 511 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA BE888491.1 - 466 Chr. - - [-] on Build
          GenBankmRNA BF304363.1 - 448 Chr. - - [-] on Build
          GenBankmRNA DB563548.1 - 2 163 Chr. 7 - 44983026 - 44983970 [-] on Build GRCh38
          GenBankmRNA BM720519.1 - 3 291 Chr. 7 - 44983133 - 44984408 [-] on Build GRCh38
          GenBankmRNA BI907654.1 - 4 448 Chr. 7 - 44983026 - 44986656 [-] on Build GRCh38
          GenBankmRNA CD523041.1 - 2 90 Chr. 7 - 44983019 - 44983896 [-] on Build GRCh38
          GenBankmRNA CN265181.1 - 2 268 Chr. 7 - 44983033 - 44984039 [-] on Build GRCh38
          GenBankmRNA BE206230.1 - 5 423 Chr. 7 - 44983229 - 44986559 [-] on Build GRCh38
          GenBankmRNA BF131634.1 - 467 Chr. - - [-] on Build
          GenBankmRNA BF306650.1 - 452 Chr. - - [-] on Build
          GenBankmRNA CF127244.1 - 166 Chr. - - [-] on Build
          GenBankmRNA AW245923.1 - 5 527 Chr. 7 - 44983229 - 44986640 [-] on Build GRCh38
          GenBankmRNA BF204739.1 - 5 521 Chr. 7 - 44983092 - 44986658 [-] on Build GRCh38
          GenBankmRNA BQ325942.1 - 5 550 Chr. 7 - 44983294 - 44986665 [-] on Build GRCh38
          GenBankmRNA BU195547.1 - 5 470 Chr. 7 - 44983030 - 44986605 [-] on Build GRCh38
          GenBankmRNA BG491747.1 - 5 544 Chr. 7 - 44983061 - 44986681 [-] on Build GRCh38
          GenBankmRNA BU502253.1 - 5 469 Chr. 7 - 44983028 - 44986604 [-] on Build GRCh38
          GenBankmRNA BM744843.1 - 5 486 Chr. 7 - 44983285 - 44986617 [-] on Build GRCh38
          GenBankmRNA DB553836.1 - 2 152 Chr. 7 - 44983026 - 44983961 [-] on Build GRCh38
          GenBankmRNA BG677227.1 - 5 459 Chr. 7 - 44983131 - 44986597 [-] on Build GRCh38
          GenBankmRNA BG829241.1 - 5 525 Chr. 7 - 44983237 - 44986619 [-] on Build GRCh38
          GenBankmRNA CD640711.1 - 5 472 Chr. 7 - 44983119 - 44986607 [-] on Build GRCh38
          GenBankmRNA BG927882.1 - 5 564 Chr. 7 - 44983170 - 44986619 [-] on Build GRCh38
          GenBankmRNA BM772795.1 - 5 486 Chr. 7 - 44983206 - 44986617 [-] on Build GRCh38
          GenBankmRNA DW440519.1 - 3 321 Chr. 7 - 44983244 - 44984422 [-] on Build GRCh38
          GenBankmRNA BM685499.1 - 5 479 Chr. 7 - 44983169 - 44986611 [-] on Build GRCh38
          GenBankmRNA AB371433.1 - 5 500 Chr. 7 - 44983029 - 44986637 [-] on Build GRCh38
          GenBankmRNA BU537835.1 - 5 530 Chr. 7 - 44983141 - 44986668 [-] on Build GRCh38
          GenBankmRNA BY996199.1 - 5 476 Chr. 7 - 44983146 - 44986608 [-] on Build GRCh38
          GenBankmRNA CD641901.1 - 4 438 Chr. 7 - 44983113 - 44986650 [-] on Build GRCh38
          GenBankmRNA AW960033.1 - 5 499 Chr. 7 - 44983173 - 44986637 [-] on Build GRCh38
          GenBankmRNA AW328480.1 - 5 437 Chr. 7 - 44983242 - 44986566 [-] on Build GRCh38
          GenBankmRNA BQ059848.1 - 351 Chr. - - [-] on Build
          GenBankmRNA BM887994.1 - 5 450 Chr. 7 - 44983268 - 44986588 [-] on Build GRCh38
          GenBankmRNA BG829322.1 - 5 521 Chr. 7 - 44983089 - 44986658 [-] on Build GRCh38
          GenBankmRNA BF308868.1 - 5 447 Chr. 7 - 44983055 - 44986586 [-] on Build GRCh38
          GenBankmRNA BG026375.1 - 308 Chr. - - [-] on Build
          GenBankmRNA AA488149.1 - 3 284 Chr. 7 - 44983085 - 44986570 [-] on Build GRCh38
          GenBankmRNA BM764510.1 - 5 486 Chr. 7 - 44983184 - 44986617 [-] on Build GRCh38
          GenBankmRNA DB560851.1 - 2 167 Chr. 7 - 44983026 - 44983974 [-] on Build GRCh38
          GenBankmRNA BE312499.1 - 5 428 Chr. 7 - 44983106 - 44986566 [-] on Build GRCh38
          GenBankmRNA BM825153.1 - 2 297 Chr. 7 - 44983026 - 44984104 [-] on Build GRCh38
          GenBankmRNA HX785737.1 - 5 511 Chr. 7 - 44983088 - 44986649 [-] on Build GRCh38
          GenBankmRNA AA306794.1 - 5 457 Chr. 7 - 44983244 - 44986595 [-] on Build GRCh38
          GenBankmRNA BG748946.1 - 5 487 Chr. 7 - 44983065 - 44986624 [-] on Build GRCh38
          GenBankmRNA BQ434731.1 - 5 500 Chr. 7 - 44983024 - 44986638 [-] on Build GRCh38
          GenBankmRNA BG548028.1 - 5 529 Chr. 7 - 44983041 - 44986665 [-] on Build GRCh38
          GenBankmRNA BU535553.1 - 5 584 Chr. 7 - 44983144 - 44986583 [-] on Build GRCh38
          GenBankmRNA CN271520.1 - 4 392 Chr. 7 - 44983122 - 44986600 [-] on Build GRCh38
          GenBankmRNA CD639046.1 - 4 438 Chr. 7 - 44983079 - 44986650 [-] on Build GRCh38
          GenBankmRNA BF317113.1 - 5 516 Chr. 7 - 44983055 - 44986653 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0