Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZFAS1 SIRNA Assays ›
          Gene Symbol:
          ZFAS1
          Gene Name:
          ZNFX1 antisense RNA 1
          Gene Aliases:
          C20orf199, HSUP1, HSUP2, NCRNA00275, ZNFX1-AS1
          Chromosome Location:
          Chr.20: 49278178 - 49289260 on Build GRCh38
          UniGene ID:
          Hs.356766
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n271359

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZFAS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003604.2 5 943
          NR_003605.1 5 624
          NR_003606.2 3 795
          NR_036658.1 4 881
          NR_036659.1 4 439
          GenBank mRNA AY339614.1 - 5 445
          AY513722.1 - 5 943
          AY513723.1 - 5 624
          BC105282.1 - 5 483
          CB137861.1 - 3 331
          CR749730.1 - 1 2096
          CR936805.1 - 4 8563
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CD385315.1 - 433 Chr. - - [-] on Build
          GenBankmRNA BG260310.1 - 305 Chr. - - [-] on Build
          GenBankmRNA CF123544.1 - 3 371 Chr. 20 - 49278639 - 49289257 [+] on Build GRCh38
          GenBankmRNA CB155482.1 - 6 547 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA DA313134.1 - 5 480 Chr. 20 - 49278642 - 49289256 [+] on Build GRCh38
          GenBankmRNA BU954336.1 - 5 482 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA AW954155.1 - 447 Chr. - - [-] on Build
          GenBankmRNA HY104293.1 - 4 416 Chr. 20 - 49278643 - 49289213 [+] on Build GRCh38
          GenBankmRNA CB151494.1 - 4 418 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU940503.1 - 5 484 Chr. 20 - 49278642 - 49289253 [+] on Build GRCh38
          GenBankmRNA BU849377.1 - 4 420 Chr. 20 - 49278642 - 49289232 [+] on Build GRCh38
          GenBankmRNA BI545004.1 - 5 488 Chr. 20 - 49278638 - 49289260 [+] on Build GRCh38
          GenBankmRNA CN295715.1 - 5 470 Chr. 20 - 49278651 - 49289260 [+] on Build GRCh38
          GenBankmRNA BG109475.1 - 5 450 Chr. 20 - 49278669 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM782711.1 - 4 417 Chr. 20 - 49278642 - 49289255 [+] on Build GRCh38
          GenBankmRNA AW953042.1 - 407 Chr. - - [-] on Build
          GenBankmRNA BE897330.1 - 4 397 Chr. 20 - 49278662 - 49289257 [+] on Build GRCh38
          GenBankmRNA CD674657.1 - 5 462 Chr. 20 - 49278659 - 49289260 [+] on Build GRCh38
          GenBankmRNA HY128835.1 - 5 478 Chr. 20 - 49278643 - 49289222 [+] on Build GRCh38
          GenBankmRNA BU599962.1 - 483 Chr. - - [-] on Build
          GenBankmRNA BM701753.1 - 5 439 Chr. 20 - 49278682 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU927839.1 - 420 Chr. - - [-] on Build
          GenBankmRNA CD248194.1 - 5 497 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BJ995977.1 - 4 417 Chr. 20 - 49278642 - 49289233 [+] on Build GRCh38
          GenBankmRNA CN353461.1 - 4 396 Chr. 20 - 49278663 - 49289231 [+] on Build GRCh38
          GenBankmRNA AA484881.1 - 4 413 Chr. 20 - 49278653 - 49289218 [+] on Build GRCh38
          GenBankmRNA BG112759.1 - 5 451 Chr. 20 - 49278670 - 49289260 [+] on Build GRCh38
          GenBankmRNA BG110424.1 - 4 449 Chr. 20 - 49278645 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU536433.1 - 4 422 Chr. 20 - 49278640 - 49289256 [+] on Build GRCh38
          GenBankmRNA EL947487.1 - 5 495 Chr. 20 - 49278667 - 49289243 [+] on Build GRCh38
          GenBankmRNA BP398633.1 - 5 459 Chr. 20 - 49278662 - 49289256 [+] on Build GRCh38
          GenBankmRNA BG530296.1 - 5 481 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA EL950104.1 - 4 397 Chr. 20 - 49278662 - 49289249 [+] on Build GRCh38
          GenBankmRNA BU170823.1 - 5 464 Chr. 20 - 49278657 - 49289256 [+] on Build GRCh38
          GenBankmRNA BU537754.1 - 4 420 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU927979.1 - 3 292 Chr. 20 - 49279156 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU943714.1 - 420 Chr. - - [-] on Build
          GenBankmRNA CN295716.1 - 5 461 Chr. 20 - 49278660 - 49289260 [+] on Build GRCh38
          GenBankmRNA CA335799.1 - 3 240 Chr. 20 - 49280544 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD173681.1 - 3 353 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CN295717.1 - 4 353 Chr. 20 - 49279155 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU541257.1 - 405 Chr. - - [-] on Build
          GenBankmRNA BP394540.1 - 4 354 Chr. 20 - 49279154 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU163963.1 - 466 Chr. - - [-] on Build
          GenBankmRNA CN353468.1 - 4 410 Chr. 20 - 49278651 - 49289257 [+] on Build GRCh38
          GenBankmRNA BG774276.1 - 5 451 Chr. 20 - 49278669 - 49289259 [+] on Build GRCh38
          GenBankmRNA BU953221.1 - 4 420 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CA337851.1 - 4 418 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA AA313204.1 - 5 452 Chr. 20 - 49278669 - 49289227 [+] on Build GRCh38
          GenBankmRNA BM738735.1 - 4 422 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA T33971.1 - 212 Chr. - - [-] on Build
          GenBankmRNA CR746822.1 - 4 358 Chr. 20 - 49279150 - 49289260 [+] on Build GRCh38
          GenBankmRNA CB124746.1 - 5 479 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD176105.1 - 5 497 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM829996.1 - 5 479 Chr. 20 - 49278642 - 49289256 [+] on Build GRCh38
          GenBankmRNA HY074449.1 - 4 416 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA BE259966.1 - 439 Chr. - - [-] on Build
          GenBankmRNA BF978934.1 - 417 Chr. - - [-] on Build
          GenBankmRNA AA149846.1 - 5 465 Chr. 20 - 49278655 - 49289250 [+] on Build GRCh38
          GenBankmRNA CN353460.1 - 4 403 Chr. 20 - 49278656 - 49289260 [+] on Build GRCh38
          GenBankmRNA BI226376.1 - 453 Chr. - - [-] on Build
          GenBankmRNA AA315192.1 - 4 420 Chr. 20 - 49278661 - 49289256 [+] on Build GRCh38
          GenBankmRNA BM714564.1 - 5 443 Chr. 20 - 49278678 - 49289251 [+] on Build GRCh38
          GenBankmRNA C14918.1 - 396 Chr. 15 - 34142268 - 34142342 [-] on Build GRCh38
          GenBankmRNA BU535035.1 - 4 420 Chr. 20 - 49278642 - 49289256 [+] on Build GRCh38
          GenBankmRNA BU537518.1 - 3 334 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BG290892.1 - 4 386 Chr. 20 - 49278672 - 49289260 [+] on Build GRCh38
          GenBankmRNA BE269507.1 - 433 Chr. - - [-] on Build
          GenBankmRNA BP395379.1 - 5 436 Chr. 20 - 49278685 - 49289259 [+] on Build GRCh38
          GenBankmRNA BU955244.1 - 3 318 Chr. 20 - 49279130 - 49289245 [+] on Build GRCh38
          GenBankmRNA CN295713.1 - 3 312 Chr. 20 - 49278669 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM309872.1 - 4 388 Chr. 20 - 49278678 - 49289243 [+] on Build GRCh38
          GenBankmRNA BM713580.1 - 4 375 Chr. 20 - 49278684 - 49289260 [+] on Build GRCh38
          GenBankmRNA CF123477.1 - 4 455 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM876538.1 - 5 449 Chr. 20 - 49278679 - 49289251 [+] on Build GRCh38
          GenBankmRNA BQ276539.1 - 483 Chr. - - [-] on Build
          GenBankmRNA CN353467.1 - 4 411 Chr. 20 - 49278648 - 49289249 [+] on Build GRCh38
          GenBankmRNA BG108581.1 - 3 291 Chr. 20 - 49278707 - 49289260 [+] on Build GRCh38
          GenBankmRNA BP398003.1 - 3 295 Chr. 20 - 49279151 - 49289256 [+] on Build GRCh38
          GenBankmRNA BQ678601.1 - 5 444 Chr. 20 - 49278677 - 49289260 [+] on Build GRCh38
          GenBankmRNA AV706790.2 - 465 Chr. - - [-] on Build
          GenBankmRNA CD175919.1 - 3 355 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CA406197.1 - 4 384 Chr. 20 - 49278673 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM785489.1 - 5 479 Chr. 20 - 49278642 - 49289251 [+] on Build GRCh38
          GenBankmRNA BE620459.1 - 5 433 Chr. 20 - 49278689 - 49289260 [+] on Build GRCh38
          GenBankmRNA AV755423.1 - 412 Chr. - - [-] on Build
          GenBankmRNA DW451942.1 - 3 348 Chr. 20 - 49278642 - 49289219 [+] on Build GRCh38
          GenBankmRNA AV703438.1 - 5 465 Chr. 20 - 49278656 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM739693.1 - 5 430 Chr. 20 - 49278690 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM465236.1 - 3 335 Chr. 20 - 49279112 - 49289260 [+] on Build GRCh38
          GenBankmRNA BP396956.1 - 3 253 Chr. 20 - 49280531 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM746639.1 - 5 503 Chr. 20 - 49278641 - 49289209 [+] on Build GRCh38
          GenBankmRNA CD299123.1 - 420 Chr. - - [-] on Build
          GenBankmRNA CD676408.1 - 4 378 Chr. 20 - 49279130 - 49289260 [+] on Build GRCh38
          GenBankmRNA BG170931.1 - 5 462 Chr. 20 - 49278658 - 49289260 [+] on Build GRCh38
          GenBankmRNA CB138604.1 - 5 478 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD048781.1 - 6 766 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA CN295714.1 - 2 271 Chr. 20 - 49279104 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM787355.1 - 4 418 Chr. 20 - 49278643 - 49289251 [+] on Build GRCh38
          GenBankmRNA W94800.1 - 455 Chr. - - [-] on Build
          GenBankmRNA CN353466.1 - 4 387 Chr. 20 - 49278672 - 49289256 [+] on Build GRCh38
          GenBankmRNA HY094146.1 - 4 420 Chr. 20 - 49278643 - 49289247 [+] on Build GRCh38
          GenBankmRNA AV744522.1 - 4 458 Chr. 20 - 49278642 - 49289232 [+] on Build GRCh38
          GenBankmRNA BE897331.1 - 4 398 Chr. 20 - 49278661 - 49289257 [+] on Build GRCh38
          GenBankmRNA BY994496.1 - 4 389 Chr. 20 - 49278670 - 49289260 [+] on Build GRCh38
          GenBankmRNA BI855943.1 - 4 385 Chr. 20 - 49278674 - 49289260 [+] on Build GRCh38
          GenBankmRNA N41712.1 - 5 474 Chr. 20 - 49278669 - 49289250 [+] on Build GRCh38
          GenBankmRNA W76265.1 - 399 Chr. - - [-] on Build
          GenBankmRNA BX463424.2 - 5 446 Chr. 20 - 49278675 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD103693.1 - 4 425 Chr. 20 - 49278640 - 49289256 [+] on Build GRCh38
          GenBankmRNA N47913.1 - 4 385 Chr. 20 - 49279123 - 49289260 [+] on Build GRCh38
          GenBankmRNA BE888655.1 - 4 382 Chr. 20 - 49278677 - 49289252 [+] on Build GRCh38
          GenBankmRNA CB140008.1 - 5 478 Chr. 20 - 49278643 - 49289236 [+] on Build GRCh38
          GenBankmRNA BM768265.1 - 5 484 Chr. 20 - 49278642 - 49289255 [+] on Build GRCh38
          GenBankmRNA AV756131.1 - 336 Chr. - - [-] on Build
          GenBankmRNA BE268167.1 - 5 430 Chr. 20 - 49278688 - 49289260 [+] on Build GRCh38
          GenBankmRNA W80527.1 - 342 Chr. - - [-] on Build
          GenBankmRNA BM476605.1 - 465 Chr. - - [-] on Build
          GenBankmRNA BM799081.1 - 3 319 Chr. 20 - 49278654 - 49289260 [+] on Build GRCh38
          GenBankmRNA CA336246.1 - 4 421 Chr. 20 - 49278638 - 49289260 [+] on Build GRCh38
          GenBankmRNA AW573582.1 - 3 247 Chr. 20 - 49280537 - 49289260 [+] on Build GRCh38
          GenBankmRNA CK005224.1 - 4 431 Chr. 20 - 49278628 - 49289256 [+] on Build GRCh38
          GenBankmRNA AA187121.1 - 4 390 Chr. 20 - 49278668 - 49289243 [+] on Build GRCh38
          GenBankmRNA H48683.1 - 4 388 Chr. 20 - 49278672 - 49289254 [+] on Build GRCh38
          GenBankmRNA BE539018.1 - 3 349 Chr. 20 - 49279104 - 49289260 [+] on Build GRCh38
          GenBankmRNA BP397712.1 - 440 Chr. - - [-] on Build
          GenBankmRNA BG112453.1 - 5 465 Chr. 20 - 49278659 - 49289242 [+] on Build GRCh38
          GenBankmRNA DW464895.1 - 3 308 Chr. 20 - 49280492 - 49289248 [+] on Build GRCh38
          GenBankmRNA BQ267294.1 - 275 Chr. - - [-] on Build
          GenBankmRNA BU600546.1 - 4 421 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM746749.1 - 5 479 Chr. 20 - 49278642 - 49289255 [+] on Build GRCh38
          GenBankmRNA BU076692.1 - 5 531 Chr. 20 - 49278271 - 49289260 [+] on Build GRCh38
          GenBankmRNA HY089030.1 - 5 479 Chr. 20 - 49278643 - 49289226 [+] on Build GRCh38
          GenBankmRNA BG034878.1 - 5 447 Chr. 20 - 49278673 - 49289260 [+] on Build GRCh38
          GenBankmRNA CB138600.1 - 5 478 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD247325.1 - 498 Chr. - - [-] on Build
          GenBankmRNA CD249562.1 - 3 349 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA AW954248.1 - 403 Chr. - - [-] on Build
          GenBankmRNA BE876452.1 - 5 447 Chr. 20 - 49278693 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU502763.1 - 465 Chr. - - [-] on Build
          GenBankmRNA CD174156.1 - 5 497 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD247470.1 - 5 496 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CR744518.1 - 4 390 Chr. 20 - 49278669 - 49289227 [+] on Build GRCh38
          GenBankmRNA HY145487.1 - 4 416 Chr. 20 - 49278643 - 49289256 [+] on Build GRCh38
          GenBankmRNA CA455555.1 - 3 254 Chr. 20 - 49280530 - 49289260 [+] on Build GRCh38
          GenBankmRNA BG831370.1 - 466 Chr. - - [-] on Build
          GenBankmRNA AJ709616.1 - 4 390 Chr. 20 - 49278669 - 49289260 [+] on Build GRCh38
          GenBankmRNA CN295712.1 - 3 323 Chr. 20 - 49278659 - 49289260 [+] on Build GRCh38
          GenBankmRNA BG621770.1 - 503 Chr. - - [-] on Build
          GenBankmRNA BP397308.1 - 443 Chr. - - [-] on Build
          GenBankmRNA BP422065.1 - 4 415 Chr. 20 - 49278666 - 49289260 [+] on Build GRCh38
          GenBankmRNA CA455477.1 - 483 Chr. - - [-] on Build
          GenBankmRNA AA306121.1 - 5 432 Chr. 20 - 49278689 - 49289250 [+] on Build GRCh38
          GenBankmRNA HY137317.1 - 4 419 Chr. 20 - 49278640 - 49289232 [+] on Build GRCh38
          GenBankmRNA CD173247.1 - 3 347 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CA395902.1 - 4 406 Chr. 20 - 49278653 - 49289256 [+] on Build GRCh38
          GenBankmRNA CD174147.1 - 3 350 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA HY150364.1 - 3 330 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA AA173594.1 - 3 251 Chr. 20 - 49280533 - 49289257 [+] on Build GRCh38
          GenBankmRNA CA337728.1 - 4 461 Chr. 20 - 49278628 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU565045.1 - 4 420 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU957909.1 - 5 482 Chr. 20 - 49278642 - 49289256 [+] on Build GRCh38
          GenBankmRNA AA400349.1 - 4 405 Chr. 20 - 49278661 - 49289230 [+] on Build GRCh38
          GenBankmRNA BI870232.1 - 5 477 Chr. 20 - 49278662 - 49289260 [+] on Build GRCh38
          GenBankmRNA D56984.1 - 3 305 Chr. 20 - 49280485 - 49289256 [+] on Build GRCh38
          GenBankmRNA CD176085.1 - 4 438 Chr. 20 - 49278640 - 49289260 [+] on Build GRCh38
          GenBankmRNA CB148257.1 - 4 417 Chr. 20 - 49278642 - 49289255 [+] on Build GRCh38
          GenBankmRNA BY797059.2 - 5 463 Chr. 20 - 49278658 - 49289257 [+] on Build GRCh38
          GenBankmRNA AV705208.1 - 4 400 Chr. 20 - 49278659 - 49289260 [+] on Build GRCh38
          GenBankmRNA BP395366.1 - 3 281 Chr. 20 - 49280503 - 49289252 [+] on Build GRCh38
          GenBankmRNA BU564699.1 - 4 420 Chr. 20 - 49278642 - 49289256 [+] on Build GRCh38
          GenBankmRNA CN295718.1 - 4 423 Chr. 20 - 49278659 - 49289260 [+] on Build GRCh38
          GenBankmRNA F29827.1 - 3 305 Chr. 20 - 49278675 - 49289209 [+] on Build GRCh38
          GenBankmRNA BG029385.1 - 408 Chr. - - [-] on Build
          GenBankmRNA AV735685.1 - 4 459 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU954247.1 - 4 421 Chr. 20 - 49278639 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU587925.1 - 2 186 Chr. 20 - 49279175 - 49289260 [+] on Build GRCh38
          GenBankmRNA AW969795.1 - 516 Chr. - - [-] on Build
          GenBankmRNA CN353463.1 - 5 470 Chr. 20 - 49278658 - 49289231 [+] on Build GRCh38
          GenBankmRNA BU963558.1 - 4 420 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD174462.1 - 5 501 Chr. 20 - 49278642 - 49289260 [+] on Build GRCh38
          GenBankmRNA CD174831.1 - 5 503 Chr. 20 - 49278643 - 49289260 [+] on Build GRCh38
          GenBankmRNA BM840527.1 - 5 480 Chr. 20 - 49278643 - 49289227 [+] on Build GRCh38
          GenBankmRNA BI199049.1 - 5 460 Chr. 20 - 49278662 - 49289260 [+] on Build GRCh38
          GenBankmRNA BX464754.2 - 5 446 Chr. 20 - 49278675 - 49289259 [+] on Build GRCh38
          GenBankmRNA CD678954.1 - 4 398 Chr. 20 - 49278661 - 49289260 [+] on Build GRCh38
          GenBankmRNA BU594883.1 - 5 482 Chr. 20 - 49278642 - 49289259 [+] on Build GRCh38
          GenBankmRNA W73489.1 - 451 Chr. - - [-] on Build
          GenBankmRNA C15238.1 - 395 Chr. 16 - 77005189 - 77005267 [-] on Build GRCh38
          GenBankmRNA AJ715322.1 - 4 390 Chr. 20 - 49278691 - 49289227 [+] on Build GRCh38
          GenBankmRNA AA429766.1 - 5 453 Chr. 20 - 49278667 - 49289211 [+] on Build GRCh38
          GenBankmRNA BU589174.1 - 3 334 Chr. 20 - 49278642 - 49289265 [+] on Build GRCh38
          GenBankmRNA BM745349.1 - 5 482 Chr. 20 - 49278640 - 49289219 [+] on Build GRCh38
          GenBankmRNA BI834636.1 - 5 468 Chr. 20 - 49278653 - 49289260 [+] on Build GRCh38
          GenBankmRNA AA102834.1 - 4 400 Chr. 20 - 49278662 - 49289260 [+] on Build GRCh38
          GenBankmRNA CN353462.1 - 4 415 Chr. 20 - 49278646 - 49289260 [+] on Build GRCh38
          GenBankmRNA W57650.1 - 405 Chr. - - [-] on Build
          GenBankmRNA HY111069.1 - 5 478 Chr. 20 - 49278643 - 49289256 [+] on Build GRCh38
          GenBankmRNA HY067680.1 - 4 422 Chr. 20 - 49278640 - 49289236 [+] on Build GRCh38
          GenBankmRNA AW965842.1 - 451 Chr. - - [-] on Build
          GenBankmRNA BM841244.1 - 5 481 Chr. 20 - 49278643 - 49289239 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0