Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NSUN5P1 SIRNA Assays ›
          Gene Symbol:
          NSUN5P1
          Gene Name:
          NOP2/Sun RNA methyltransferase family member 5 pseudogene 1
          Gene Aliases:
          NSUN5B, WBSCR20B
          Chromosome Location:
          Chr.7: 75410322 - 75416792 on Build GRCh38
          UniGene ID:
          Hs.436034
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n271652

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NSUN5P1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033322.3 9 1288
          NR_104013.1 7 1323
          GenBank mRNA AF416610.1 - 8 1152
          AK094312.1 - 3 2266
          AK126486.1 - 5 4240
          AK128374.1 - 3 2478
          BC093978.1 - 1 248 7939823
          BC107112.1 - 2 342 40022630
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AL519766.3 - 5 691 Chr. 7 - 75410382 - 75416435 [+] on Build GRCh38
          GenBankmRNA BM563705.1 - 361 Chr. - - [-] on Build
          GenBankmRNA BM922808.1 - 1 273 Chr. 7 - 75415879 - 75416972 [+] on Build GRCh38
          GenBankmRNA CN278913.1 - 1 198 Chr. 7 - 75415961 - 75416787 [+] on Build GRCh38
          GenBankmRNA CN398823.1 - 1 424 Chr. 7 - 75415740 - 75416443 [+] on Build GRCh38
          GenBankmRNA BQ436510.1 - 5 413 Chr. 7 - 75410401 - 75416654 [+] on Build GRCh38
          GenBankmRNA CD676348.1 - 3 287 Chr. 7 - 75415591 - 75416667 [+] on Build GRCh38
          GenBankmRNA AW957437.1 - 1 348 Chr. 7 - 75415804 - 75416515 [+] on Build GRCh38
          GenBankmRNA AV691151.1 - 1 206 Chr. 7 - 75415946 - 75416488 [+] on Build GRCh38
          GenBankmRNA DB522712.2 - 1 156 Chr. 7 - 75416001 - 75416785 [+] on Build GRCh38
          GenBankmRNA BG006063.1 - 1 257 Chr. 7 - 75415897 - 75416397 [+] on Build GRCh38
          GenBankmRNA CK003229.1 - 1 305 Chr. 7 - 75415849 - 75416605 [+] on Build GRCh38
          GenBankmRNA AV692842.1 - 1 206 Chr. 7 - 75415946 - 75416443 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA base methylation
          RNA binding
          RNA methyltransferase activity
          S-adenosylmethionine-dependent methyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0