Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGAP2-AS1 SIRNA Assays ›
          Gene Symbol:
          AGAP2-AS1
          Gene Name:
          AGAP2 antisense RNA 1
          Gene Aliases:
          PUNISHER
          Chromosome Location:
          Chr.12: 57726240 - 57728356 on Build GRCh38
          UniGene ID:
          Hs.656080
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n272082

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AGAP2-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027032.1 2 523
          GenBank mRNA AK301303.1 - 2 497
          BC039697.1 - 2 442
          CN313469.1 - 2 523
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM459742.1 - 2 447 Chr. 12 - 57726319 - 57727715 [+] on Build GRCh38
          GenBankmRNA BQ668531.1 - 459 Chr. - - [-] on Build
          GenBankmRNA BI117754.1 - 2 507 Chr. 12 - 57726279 - 57727600 [+] on Build GRCh38
          GenBankmRNA BQ668985.1 - 2 459 Chr. 12 - 57726304 - 57727673 [+] on Build GRCh38
          GenBankmRNA BG327113.1 - 2 453 Chr. 12 - 57726310 - 57727409 [+] on Build GRCh38
          GenBankmRNA BM911481.1 - 488 Chr. - - [-] on Build
          GenBankmRNA BM561078.1 - 490 Chr. - - [-] on Build
          GenBankmRNA BQ670088.1 - 2 458 Chr. 12 - 57726304 - 57727709 [+] on Build GRCh38
          GenBankmRNA BG423979.1 - 2 478 Chr. 12 - 57726286 - 57727467 [+] on Build GRCh38
          GenBankmRNA BI756796.1 - 2 452 Chr. 12 - 57726310 - 57727815 [+] on Build GRCh38
          GenBankmRNA BX387184.2 - 2 507 Chr. 12 - 57726280 - 57727700 [+] on Build GRCh38
          GenBankmRNA BM556361.1 - 2 447 Chr. 12 - 57726319 - 57727875 [+] on Build GRCh38
          GenBankmRNA BI755479.1 - 2 449 Chr. 12 - 57726316 - 57727714 [+] on Build GRCh38
          GenBankmRNA BM559643.1 - 2 461 Chr. 12 - 57726301 - 57727840 [+] on Build GRCh38
          GenBankmRNA AL546231.3 - 2 474 Chr. 12 - 57726288 - 57727861 [+] on Build GRCh38
          GenBankmRNA BG491431.1 - 2 464 Chr. 12 - 57726299 - 57727635 [+] on Build GRCh38
          GenBankmRNA BM808141.1 - 2 489 Chr. 12 - 57726274 - 57727809 [+] on Build GRCh38
          GenBankmRNA AL525564.3 - 2 485 Chr. 12 - 57726277 - 57727774 [+] on Build GRCh38
          GenBankmRNA BM009830.1 - 2 493 Chr. 12 - 57726271 - 57727657 [+] on Build GRCh38
          GenBankmRNA BU173742.1 - 2 459 Chr. 12 - 57726304 - 57727725 [+] on Build GRCh38
          GenBankmRNA BI914975.1 - 445 Chr. - - [-] on Build
          GenBankmRNA BE296133.1 - 2 432 Chr. 12 - 57726335 - 57727503 [+] on Build GRCh38
          GenBankmRNA BQ670297.1 - 2 482 Chr. 12 - 57726307 - 57727757 [+] on Build GRCh38
          GenBankmRNA BE264977.1 - 2 434 Chr. 12 - 57726330 - 57727591 [+] on Build GRCh38
          GenBankmRNA CB141359.1 - 1 247 Chr. 12 - 57727102 - 57727730 [+] on Build GRCh38
          GenBankmRNA AL524680.3 - 2 459 Chr. 12 - 57726303 - 57727907 [+] on Build GRCh38
          GenBankmRNA DC392337.1 - 2 498 Chr. 12 - 57726266 - 57727405 [+] on Build GRCh38
          GenBankmRNA BE251649.1 - 2 420 Chr. 12 - 57726345 - 57727589 [+] on Build GRCh38
          GenBankmRNA AW245733.1 - 2 469 Chr. 12 - 57726311 - 57727462 [+] on Build GRCh38
          GenBankmRNA BX353593.2 - 2 506 Chr. 12 - 57726280 - 57727874 [+] on Build GRCh38
          GenBankmRNA CN313468.1 - 2 460 Chr. 12 - 57726303 - 57727515 [+] on Build GRCh38
          GenBankmRNA BF309969.1 - 447 Chr. - - [-] on Build
          GenBankmRNA BM914685.1 - 461 Chr. - - [-] on Build
          GenBankmRNA BQ675318.1 - 2 480 Chr. 12 - 57726282 - 57727721 [+] on Build GRCh38
          GenBankmRNA BM009331.1 - 2 493 Chr. 12 - 57726271 - 57727739 [+] on Build GRCh38
          GenBankmRNA BM556521.1 - 2 519 Chr. 12 - 57726271 - 57727724 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5l6s2:80/100.66.78.182:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0