Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGAP2-AS1 SIRNA Assays ›
          Gene Symbol:
          AGAP2-AS1
          Gene Name:
          AGAP2 antisense RNA 1
          Gene Aliases:
          PUNISHER
          Chromosome Location:
          Chr.12: 57726240 - 57728356 on Build GRCh38
          UniGene ID:
          Hs.656080
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n272083

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AGAP2-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027032.1 2 271
          GenBank mRNA AK301303.1 - 2 245
          BC039697.1 - 2 190
          CN313469.1 - 2 271
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM459742.1 - 2 195 Chr. 12 - 57726319 - 57727715 [+] on Build GRCh38
          GenBankmRNA BQ668531.1 - 207 Chr. - - [-] on Build
          GenBankmRNA BI117754.1 - 2 255 Chr. 12 - 57726279 - 57727600 [+] on Build GRCh38
          GenBankmRNA BQ668985.1 - 2 207 Chr. 12 - 57726304 - 57727673 [+] on Build GRCh38
          GenBankmRNA BG469657.1 - 228 Chr. - - [-] on Build
          GenBankmRNA BG327113.1 - 2 201 Chr. 12 - 57726310 - 57727409 [+] on Build GRCh38
          GenBankmRNA BM911481.1 - 236 Chr. - - [-] on Build
          GenBankmRNA BM561078.1 - 238 Chr. - - [-] on Build
          GenBankmRNA BQ670088.1 - 2 206 Chr. 12 - 57726304 - 57727709 [+] on Build GRCh38
          GenBankmRNA BG423979.1 - 2 226 Chr. 12 - 57726286 - 57727467 [+] on Build GRCh38
          GenBankmRNA BI756796.1 - 2 201 Chr. 12 - 57726310 - 57727815 [+] on Build GRCh38
          GenBankmRNA BX387184.2 - 2 255 Chr. 12 - 57726280 - 57727700 [+] on Build GRCh38
          GenBankmRNA BM556361.1 - 2 195 Chr. 12 - 57726319 - 57727875 [+] on Build GRCh38
          GenBankmRNA BI755479.1 - 2 196 Chr. 12 - 57726316 - 57727714 [+] on Build GRCh38
          GenBankmRNA HY021756.1 - 2 236 Chr. 12 - 57726275 - 57727335 [+] on Build GRCh38
          GenBankmRNA AL546231.3 - 2 222 Chr. 12 - 57726288 - 57727861 [+] on Build GRCh38
          GenBankmRNA BM559643.1 - 2 209 Chr. 12 - 57726301 - 57727840 [+] on Build GRCh38
          GenBankmRNA BG491431.1 - 2 213 Chr. 12 - 57726299 - 57727635 [+] on Build GRCh38
          GenBankmRNA BM808141.1 - 2 237 Chr. 12 - 57726274 - 57727809 [+] on Build GRCh38
          GenBankmRNA AL525564.3 - 2 233 Chr. 12 - 57726277 - 57727774 [+] on Build GRCh38
          GenBankmRNA BM009830.1 - 2 241 Chr. 12 - 57726271 - 57727657 [+] on Build GRCh38
          GenBankmRNA BI914975.1 - 193 Chr. - - [-] on Build
          GenBankmRNA BU173742.1 - 2 207 Chr. 12 - 57726304 - 57727725 [+] on Build GRCh38
          GenBankmRNA BQ670297.1 - 2 230 Chr. 12 - 57726307 - 57727757 [+] on Build GRCh38
          GenBankmRNA BE264977.1 - 2 182 Chr. 12 - 57726330 - 57727591 [+] on Build GRCh38
          GenBankmRNA AL524680.3 - 2 207 Chr. 12 - 57726303 - 57727907 [+] on Build GRCh38
          GenBankmRNA BE251649.1 - 2 166 Chr. 12 - 57726345 - 57727589 [+] on Build GRCh38
          GenBankmRNA DC392337.1 - 2 246 Chr. 12 - 57726266 - 57727405 [+] on Build GRCh38
          GenBankmRNA BX353593.2 - 2 254 Chr. 12 - 57726280 - 57727874 [+] on Build GRCh38
          GenBankmRNA AW245733.1 - 2 217 Chr. 12 - 57726311 - 57727462 [+] on Build GRCh38
          GenBankmRNA BF309969.1 - 196 Chr. - - [-] on Build
          GenBankmRNA CN313468.1 - 2 208 Chr. 12 - 57726303 - 57727515 [+] on Build GRCh38
          GenBankmRNA BM914685.1 - 209 Chr. - - [-] on Build
          GenBankmRNA BQ675318.1 - 2 228 Chr. 12 - 57726282 - 57727721 [+] on Build GRCh38
          GenBankmRNA BM556521.1 - 2 267 Chr. 12 - 57726271 - 57727724 [+] on Build GRCh38
          GenBankmRNA BM009331.1 - 2 243 Chr. 12 - 57726271 - 57727739 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0