Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other COPB2 SIRNA Assays ›
          Gene Symbol:
          COPB2
          Gene Name:
          coatomer protein complex subunit beta 2
          Gene Aliases:
          beta'-COP
          Chromosome Location:
          Chr.3: 139355595 - 139389680 on Build GRCh38
          UniGene ID:
          Hs.681616
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n272183

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: COPB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_023350.1 1 140
          GenBank mRNA DC385126.1 - 1 99
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DC352604.1 - 1 40 Chr. 3 - 139378111 - 139389580 [-] on Build GRCh38
          GenBankmRNA DC298303.1 - 1 99 Chr. 3 - 139378148 - 139389639 [-] on Build GRCh38
          GenBankmRNA DC302185.1 - 1 103 Chr. 3 - 139378176 - 139389643 [-] on Build GRCh38
          GenBankmRNA DC309521.1 - 1 140 Chr. 3 - 139378159 - 139389680 [-] on Build GRCh38
          GenBankmRNA DC407794.1 - 1 76 Chr. 3 - 139378117 - 139389616 [-] on Build GRCh38
          GenBankmRNA DC346442.1 - 1 99 Chr. 3 - 139378155 - 139389639 [-] on Build GRCh38
          GenBankmRNA DC362516.1 - 1 117 Chr. 3 - 139378151 - 139389657 [-] on Build GRCh38
          GenBankmRNA BX456895.2 - 1 136 Chr. 3 - 139379413 - 139389651 [-] on Build GRCh38
          GenBankmRNA CN281888.1 - 1 77 Chr. 3 - 139379430 - 139389616 [-] on Build GRCh38
          GenBankmRNA DC363335.1 - 1 103 Chr. 3 - 139378168 - 139389643 [-] on Build GRCh38
          GenBankmRNA DC330801.1 - 1 108 Chr. 3 - 139378142 - 139389648 [-] on Build GRCh38
          GenBankmRNA DC369721.1 - 1 76 Chr. 3 - 139378125 - 139389616 [-] on Build GRCh38
          GenBankmRNA DC353591.1 - 1 109 Chr. 3 - 139378141 - 139389649 [-] on Build GRCh38
          GenBankmRNA DC321829.1 - 1 43 Chr. 3 - 139378096 - 139389583 [-] on Build GRCh38
          GenBankmRNA BQ229979.1 - 1 96 Chr. 3 - 139375476 - 139389636 [-] on Build GRCh38
          GenBankmRNA DC370020.1 - 1 117 Chr. 3 - 139378162 - 139389657 [-] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          ER to Golgi vesicle-mediated transport
          retrograde vesicle-mediated transport, Golgi to ER
          intra-Golgi vesicle-mediated transport
          toxin transport
          structural molecule activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0