Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MEG3 SIRNA Assays ›
          Gene Symbol:
          MEG3
          Gene Name:
          maternally expressed 3 (non-protein coding)
          Gene Aliases:
          FP504, GTL2, LINC00023, NCRNA00023, PRO0518, PRO2160, onco-lncRNA-83, prebp1
          Chromosome Location:
          Chr.14: 100826108 - 100861026 on Build GRCh38
          UniGene ID:
          Hs.654863
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n272553

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MEG3
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002766.2
          3 351
          NR_003530.2
          3 351
          NR_003531.3
          3 327
          NR_033358.1
          3 351
          NR_033359.1
          3 351
          NR_033360.1
          3 351
          NR_046464.1
          3 351
          NR_046465.2
          3 351
          NR_046466.1
          3 351
          NR_046467.1
          3 351
          NR_046469.1
          3 351
          NR_046470.2
          3 351
          NR_046471.1
          3 351
          NR_046472.1
          3 351
          NR_046473.1
          3 351
          GenBank mRNA AB032607.1 -
          3 340
          AF052114.1 -
          1 93
          AY314975.1 - LIT3321
          MEG3
          3 327
          BC023543.2 -
          3 326
          DB449068.1 -
          3 350
          GQ183494.1 -
          3 343
          GQ183495.1 -
          3 343
          GQ183496.1 -
          3 343
          GQ183497.1 -
          3 343
          GQ183498.1 -
          3 343
          GQ183499.1 -
          3 343
          GQ183500.1 -
          3 343
          GQ183501.1 -
          3 343
          GQ183502.1 -
          3 343
          GQ183503.1 -
          3 343
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GENCODE ENST00000451743.6 - 3 333 Chr. 14 - 100826126 - 100861031 [+] on Build GRCh38
          GENCODE ENST00000556736.5 - 3 333 Chr. 14 - 100826126 - 100861008 [+] on Build GRCh38
          GENCODE ENST00000452120.6 - 3 326 Chr. 14 - 100826133 - 100833328 [+] on Build GRCh38
          GENCODE ENST00000554639.5 - 3 327 Chr. 14 - 100826132 - 100861026 [+] on Build GRCh38
          GENCODE ENST00000398518.6 - 3 332 Chr. 14 - 100826127 - 100860994 [+] on Build GRCh38
          GENCODE ENST00000412736.6 - 3 311 Chr. 14 - 100826148 - 100860977 [+] on Build GRCh38
          GENCODE ENST00000423456.5 - 3 333 Chr. 14 - 100826126 - 100860970 [+] on Build GRCh38
          GENCODE ENST00000429159.6 - 3 351 Chr. 14 - 100826108 - 100861023 [+] on Build GRCh38
          GENCODE ENST00000455286.1 - 3 358 Chr. 14 - 100827411 - 100829306 [+] on Build GRCh38
          GENCODE ENST00000521404.5 - 3 333 Chr. 14 - 100826126 - 100860989 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0