Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPL10 SIRNA Assays ›
          Gene Symbol:
          RPL10
          Gene Name:
          ribosomal protein L10
          Gene Aliases:
          AUTSX5, DXS648, DXS648E, L10, NOV, QM
          Chromosome Location:
          Chr.X: 154398065 - 154402339 on Build GRCh38
          UniGene ID:
          Hs.534404
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n273489

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RPL10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AA187603.1 - 2 319
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA HY253991.1 - 1 346 Chr. 5 - 168616584 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY292846.1 - 1 344 Chr. 5 - 168616621 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY253044.1 - 1 346 Chr. 5 - 168616611 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY348364.1 - 1 345 Chr. 5 - 168616665 - 168617067 [-] on Build GRCh38
          GenBankmRNA HY256786.1 - 1 344 Chr. 5 - 168616558 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY257175.1 - 1 345 Chr. 5 - 168616570 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY274544.1 - 1 344 Chr. 19 - 12643203 - 12643653 [+] on Build GRCh38
          GenBankmRNA HY272818.1 - 1 347 Chr. 5 - 168616649 - 168617069 [-] on Build GRCh38
          GenBankmRNA HY289225.1 - 1 345 Chr. 5 - 168616654 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY256852.1 - 1 346 Chr. 5 - 168616602 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY265927.1 - 1 342 Chr. 5 - 168616578 - 168617067 [-] on Build GRCh38
          GenBankmRNA AY927536.3 - 2 339 Chr. X - 154399883 - 154403497 [-] on Build GRCh38
          GenBankmRNA HY290915.1 - 1 344 Chr. 5 - 168616625 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY279150.1 - 2 346 Chr. X - 154400442 - 154400913 [-] on Build GRCh38
          GenBankmRNA HY275660.1 - 1 346 Chr. 5 - 168616617 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY288832.1 - 1 343 Chr. 5 - 168616645 - 168617067 [-] on Build GRCh38
          GenBankmRNA HY277578.1 - 1 344 Chr. 5 - 168616643 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY370884.1 - 1 346 Chr. 5 - 168616634 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY270299.1 - 1 346 Chr. 5 - 168616632 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY298555.1 - 1 343 Chr. 5 - 168616601 - 168617067 [-] on Build GRCh38
          GenBankmRNA HY266266.1 - 1 346 Chr. 5 - 168616583 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY272479.1 - 1 343 Chr. 5 - 168616656 - 168617067 [-] on Build GRCh38
          GenBankmRNA HY256774.1 - 1 346 Chr. 5 - 168616553 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY248058.1 - 1 344 Chr. 5 - 168616630 - 168617067 [-] on Build GRCh38
          GenBankmRNA HY245517.1 - 1 345 Chr. 5 - 168616547 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY183571.1 - 1 346 Chr. 5 - 168616622 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY275991.1 - 1 343 Chr. 5 - 168616656 - 168617067 [-] on Build GRCh38
          GenBankmRNA HY295044.1 - 1 344 Chr. 5 - 168616620 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY256904.1 - 1 346 Chr. 5 - 168616559 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY286029.1 - 1 348 Chr. 19 - 12643199 - 12643660 [+] on Build GRCh38
          GenBankmRNA HY279766.1 - 1 344 Chr. 5 - 168616659 - 168617067 [-] on Build GRCh38
          GenBankmRNA HY322604.1 - 1 345 Chr. 5 - 168616634 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY261912.1 - 1 344 Chr. 5 - 168616594 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY095117.1 - 1 360 Chr. 5 - 168616640 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY285256.1 - 1 344 Chr. 5 - 168616627 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY292306.1 - 2 346 Chr. X - 154400402 - 154400913 [-] on Build GRCh38
          GenBankmRNA HY322826.1 - 1 347 Chr. 19 - 12643200 - 12643667 [+] on Build GRCh38
          GenBankmRNA HY254159.1 - 1 376 Chr. 19 - 12643171 - 12643662 [+] on Build GRCh38
          GenBankmRNA HY296389.1 - 1 344 Chr. 5 - 168616632 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY290347.1 - 1 343 Chr. 5 - 168616647 - 168617067 [-] on Build GRCh38
          GenBankmRNA HY269042.1 - 1 344 Chr. 5 - 168616666 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY259788.1 - 1 347 Chr. 5 - 168616575 - 168617069 [-] on Build GRCh38
          GenBankmRNA HY269849.1 - 1 343 Chr. 5 - 168616645 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY249955.1 - 1 346 Chr. 5 - 168616568 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY277912.1 - 1 348 Chr. 19 - 12643201 - 12643641 [+] on Build GRCh38
          GenBankmRNA HY275359.1 - 1 344 Chr. 5 - 168616612 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY256939.1 - 1 344 Chr. 5 - 168616566 - 168617067 [-] on Build GRCh38
          GenBankmRNA HY337617.1 - 1 344 Chr. 5 - 168616558 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY247371.1 - 1 344 Chr. 5 - 168616562 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY272777.1 - 1 344 Chr. 5 - 168616658 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY256409.1 - 1 346 Chr. 5 - 168616598 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY251886.1 - 1 344 Chr. 5 - 168616551 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY259409.1 - 1 346 Chr. 5 - 168616581 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY263275.1 - 1 345 Chr. 5 - 168616597 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY290015.1 - 1 349 Chr. 19 - 12643199 - 12643643 [+] on Build GRCh38
          GenBankmRNA HY280020.1 - 1 346 Chr. 5 - 168616603 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY263641.1 - 1 346 Chr. 5 - 168616614 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY292986.1 - 1 344 Chr. 5 - 168616614 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY294491.1 - 1 344 Chr. 5 - 168616643 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY280211.1 - 1 346 Chr. 5 - 168616617 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY301068.1 - 1 344 Chr. 5 - 168616640 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY234374.1 - 1 343 Chr. 5 - 168616619 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY327669.1 - 1 345 Chr. 5 - 168616647 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY279188.1 - 1 345 Chr. 5 - 168616664 - 168617068 [-] on Build GRCh38
          GenBankmRNA HY271271.1 - 1 420 Chr. X - 154400460 - 154400913 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0