Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other JPX SIRNA Assays ›
          Gene Symbol:
          JPX
          Gene Name:
          JPX transcript, XIST activator (non-protein coding)
          Gene Aliases:
          DCBALD06, ENOX, LINC00183, NCRNA00183
          Chromosome Location:
          Chr.X: 73944324 - 74070384 on Build GRCh38
          UniGene ID:
          Hs.648316
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n273720

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: JPX
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK056172.1 -
          3 471
          AV714079.1 - LIT1011
          Jpx
          474
          BP871721.1 -
          3 439
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BG504771.1 - 3 467 Chr. X - 73944346 - 73947528 [+] on Build GRCh38
          GenBankmRNA DA979750.1 - 3 461 Chr. X - 73944351 - 73947352 [+] on Build GRCh38
          GenBankmRNA BU164379.1 - 3 464 Chr. X - 73944349 - 73998832 [+] on Build GRCh38
          GenBankmRNA BP871836.1 - 3 467 Chr. X - 73944346 - 73947297 [+] on Build GRCh38
          GenBankmRNA AW963219.1 - 461 Chr. - - [-] on Build
          GenBankmRNA DA427047.1 - 3 487 Chr. X - 73944326 - 73947342 [+] on Build GRCh38
          GenBankmRNA DB240432.1 - 3 467 Chr. X - 73944346 - 73947385 [+] on Build GRCh38
          GenBankmRNA DA841100.1 - 3 468 Chr. X - 73944345 - 73947361 [+] on Build GRCh38
          GenBankmRNA AL708895.1 - 3 476 Chr. X - 73944338 - 73947333 [+] on Build GRCh38
          GenBankmRNA DA731241.1 - 3 487 Chr. X - 73944326 - 73998845 [+] on Build GRCh38
          GenBankmRNA BU537716.1 - 3 483 Chr. X - 73944333 - 73947334 [+] on Build GRCh38
          GenBankmRNA DB255033.1 - 3 476 Chr. X - 73944339 - 73947366 [+] on Build GRCh38
          GenBankmRNA DA711535.1 - 3 472 Chr. X - 73944342 - 73947373 [+] on Build GRCh38
          GenBankmRNA DA700446.1 - 3 472 Chr. X - 73944342 - 73947318 [+] on Build GRCh38
          GenBankmRNA BM810643.1 - 444 Chr. - - [-] on Build
          GenBankmRNA BP871862.1 - 3 473 Chr. X - 73944340 - 73947374 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-6h858:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0