Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYO15B SIRNA Assays ›
          Gene Symbol:
          MYO15B
          Gene Name:
          myosin XVB
          Gene Aliases:
          MYO15BP
          Chromosome Location:
          Chr.17: 75587545 - 75626850 on Build GRCh38
          UniGene ID:
          Hs.390817
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n274575

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYO15B
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001309242.1 NP_001296171.1
          60, 61 8822
          XM_017025120.1 XP_016880609.1
          62, 63 8529
          XM_017025121.1 XP_016880610.1
          62, 63 8514
          XM_017025122.1 XP_016880611.1
          62, 63 8493
          XM_017025123.1 XP_016880612.1
          62, 63 8465
          XM_017025124.1 XP_016880613.1
          61, 62 8403
          XM_017025125.1 XP_016880614.1
          60, 61 8252
          XM_017025126.1 XP_016880615.1
          57, 58 7994
          XM_017025127.1 XP_016880616.1
          60, 61 7837
          XM_017025128.1 XP_016880617.1
          61, 62 7583
          XM_017025129.1 XP_016880618.1
          60, 61 6815
          XM_017025130.1 XP_016880619.1
          59, 60 6771
          XM_017025131.1 XP_016880620.1
          58, 59 7210
          XM_017025132.1 XP_016880621.1
          57, 58 7151
          XM_017025133.1 XP_016880622.1
          55, 56 6676
          XM_017025134.1 XP_016880623.1
          56, 57 7037
          XM_017025135.1 XP_016880624.1
          50, 51 5693
          XM_017025136.1 XP_016880625.1
          50, 51 6042
          XM_017025137.1 XP_016880626.1
          47, 48 5587
          XM_017025138.1 XP_016880627.1
          42, 43 5083
          XM_017025139.1 XP_016880628.1
          43, 44 4884
          XM_017025140.1 XP_016880629.1
          40, 41 4697
          XM_017025143.1 XP_016880632.1
          35, 36 3974
          XM_017025144.1 XP_016880633.1
          56, 57 8457
          GenBank mRNA AF418292.1 -
          35, 36 4432
          AF418293.1 -
          36, 37 4038
          AK096869.1 -
          12, 13 1276
          AL713730.1 -
          11, 12 1031
          BC027875.1 - LIT3316
          MYO15BP
          12, 13 1526
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BG680906.1 - 7, 8 757 Chr. 17 - 75623997 - 75625956 [+] on Build GRCh38
          GenBankmRNA BP426098.1 - 1, 2 19 Chr. 17 - 75625640 - 75626229 [+] on Build GRCh38
          GenBankmRNA CN259055.1 - 3, 4 335 Chr. 17 - 75624826 - 75626501 [+] on Build GRCh38
          GenBankmRNA BU150940.1 - 5, 6 510 Chr. 17 - 75624429 - 75626440 [+] on Build GRCh38
          GenBankmRNA BG426026.1 - 409 Chr. - - [-] on Build
          GenBankmRNA AV683102.1 - 3, 4 373 Chr. 17 - 75624788 - 75626229 [+] on Build GRCh38
          GenBankmRNA BG953175.1 - 2, 3 164 Chr. 17 - 75556495 - 75626119 [+] on Build GRCh38
          GenBankmRNA T86223.1 - 34 Chr. - - [-] on Build

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0