Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYO15B SIRNA Assays ›
          Gene Symbol:
          MYO15B
          Gene Name:
          myosin XVB
          Gene Aliases:
          MYO15BP
          Chromosome Location:
          Chr.17: 75587545 - 75626850 on Build GRCh38
          UniGene ID:
          Hs.390817
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n274576

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYO15B
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001309242.1 NP_001296171.1
          55, 56 8250
          XM_017025120.1 XP_016880609.1
          57, 58 7957
          XM_017025121.1 XP_016880610.1
          57, 58 7942
          XM_017025122.1 XP_016880611.1
          57, 58 7921
          XM_017025123.1 XP_016880612.1
          57, 58 7893
          XM_017025124.1 XP_016880613.1
          56, 57 7831
          XM_017025125.1 XP_016880614.1
          55, 56 7680
          XM_017025126.1 XP_016880615.1
          52, 53 7422
          XM_017025127.1 XP_016880616.1
          55, 56 7265
          XM_017025128.1 XP_016880617.1
          56, 57 7011
          XM_017025129.1 XP_016880618.1
          55, 56 6243
          XM_017025130.1 XP_016880619.1
          54, 55 6199
          XM_017025131.1 XP_016880620.1
          53, 54 6638
          XM_017025132.1 XP_016880621.1
          52, 53 6579
          XM_017025133.1 XP_016880622.1
          50, 51 6104
          XM_017025134.1 XP_016880623.1
          51, 52 6465
          XM_017025135.1 XP_016880624.1
          45, 46 5121
          XM_017025136.1 XP_016880625.1
          45, 46 5470
          XM_017025137.1 XP_016880626.1
          42, 43 5015
          XM_017025138.1 XP_016880627.1
          37, 38 4511
          XM_017025139.1 XP_016880628.1
          38, 39 4312
          XM_017025140.1 XP_016880629.1
          35, 36 4125
          XM_017025143.1 XP_016880632.1
          30, 31 3402
          XM_017025144.1 XP_016880633.1
          51, 52 7885
          GenBank mRNA AF418292.1 -
          30, 31 3860
          AF418293.1 -
          31, 32 3466
          AK026339.1 -
          7, 8 565
          AK096869.1 -
          7, 8 704
          AL713730.1 -
          6, 7 459
          BC027875.1 - LIT3316
          MYO15BP
          7, 8 954
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AW753302.1 - 2, 3 131 Chr. 17 - 75624028 - 75625581 [+] on Build GRCh38
          GenBankmRNA BE145755.1 - 2, 3 133 Chr. 17 - 75624023 - 75624571 [+] on Build GRCh38
          GenBankmRNA CV029543.1 - 7, 8 537 Chr. 17 - 75621133 - 75624449 [+] on Build GRCh38
          GenBankmRNA BG680906.1 - 2, 3 185 Chr. 17 - 75623997 - 75625956 [+] on Build GRCh38
          GenBankmRNA CD579419.2 - 383 Chr. - - [-] on Build
          GenBankmRNA BM720779.1 - 6, 7 460 Chr. 17 - 75621379 - 75624849 [+] on Build GRCh38
          GenBankmRNA DN997131.1 - 685 Chr. 17 - 75620992 - 75624269 [+] on Build GRCh38
          GenBankmRNA BG912704.1 - 7, 8 529 Chr. 17 - 75621075 - 75624899 [+] on Build GRCh38
          GenBankmRNA BI766411.1 - 3, 4 234 Chr. 17 - 75623828 - 75625879 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-jmx4f:80/100.66.76.53:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0