Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NLGN1 SIRNA Assays ›
Gene Symbol:
NLGN1
Gene Name:
neuroligin 1
Gene Aliases:
NL1
Chromosome Location:
Chr.3: 173395899 - 174291287 on Build GRCh38
UniGene ID:
Hs.478289
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n277906
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NLGN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_005247234.2
XP_005247291.1
6
7123
XM_005247235.3
XP_005247292.1
6
7319
XM_005247236.2
XP_005247293.1
6
7096
XM_005247237.3
XP_005247294.1
5
7617
XM_006713540.3
XP_006713603.1
7
7379
XM_011512551.2
XP_011510853.1
8
7230
XM_011512552.2
XP_011510854.1
7
7666
XM_011512553.2
XP_011510855.1
4
6019
XM_011512554.2
XP_011510856.1
4
5979
XM_017005888.1
XP_016861377.1
7
7527
XM_017005889.1
XP_016861378.1
7
7527
XM_017005890.1
XP_016861379.1
7
7183
XM_017005891.1
XP_016861380.1
7
7379
XM_017005892.1
XP_016861381.1
7
7681
XM_017005893.1
XP_016861382.1
8
7793
XM_017005894.1
XP_016861383.1
7
9311
XM_017005895.1
XP_016861384.1
7
13484
XM_017005896.1
XP_016861385.1
7
6965
XM_017005897.1
XP_016861386.1
7
7500
XM_017005898.1
XP_016861387.1
6
7467
XM_017005899.1
XP_016861388.1
6
7606
XM_017005900.1
XP_016861389.1
7
7112
XM_017005901.1
XP_016861390.1
6
7168
XM_017005902.1
XP_016861391.1
7
7143
XM_017005903.1
XP_016861392.1
6
7292
XM_017005904.1
XP_016861393.1
6
7579
XM_017005905.1
XP_016861394.1
4
7310
GenBank mRNA
AK129874.1
-
1
815
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
Transcript Location
GenBankmRNA
CD106220.1
-
1
97
Chr. 3 - 174285254 - 174286103 [+] on Build GRCh38
GenBankmRNA
BM453850.1
-
1
165
Chr. 3 - 174285186 - 174286046 [+] on Build GRCh38
Back To Top
More Information
Panther Classification:
Molecular Function -
cell adhesion molecule
Gene Ontology Categories:
Function(s)
Process(es)
regulation of respiratory gaseous exchange by neurological system process
protein targeting
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
neuron cell-cell adhesion
nervous system development
synapse assembly
positive regulation of circadian sleep/wake cycle, wakefulness
synaptic vesicle targeting
calcium-dependent cell-cell adhesion via plasma membrane cell adhesion molecules
neuronal signal transduction
neuron projection development
positive regulation of synaptic transmission, GABAergic
protein localization to synapse
establishment of protein localization
regulation of neuron differentiation
rhythmic process
cytoskeletal matrix organization at active zone
protein homooligomerization
protein heterotetramerization
positive regulation of filopodium assembly
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
long-term synaptic potentiation
positive regulation of dendritic spine development
negative regulation of dendritic spine morphogenesis
terminal button organization
synaptic vesicle clustering
postsynaptic membrane assembly
presynaptic membrane assembly
AMPA glutamate receptor clustering
NMDA glutamate receptor clustering
neurexin clustering involved in presynaptic membrane assembly
postsynaptic density protein 95 clustering
receptor localization to synapse
positive regulation of ruffle assembly
positive regulation of synaptic vesicle endocytosis
positive regulation of protein localization to synapse
positive regulation of intracellular signal transduction
positive regulation of synaptic vesicle exocytosis
regulation of N-methyl-D-aspartate selective glutamate receptor activity
regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
positive regulation of excitatory postsynaptic potential
positive regulation of synaptic vesicle clustering
receptor activity
PDZ domain binding
neurexin family protein binding
protein dimerization activity
cell adhesion molecule binding
carboxylic ester hydrolase activity
scaffold protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0