Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CEP85L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042475.2 | NP_001035940.1 | 13 | 7348 | ||
NM_001178035.1 | NP_001171506.1 | 14 | 6836 | |||
XM_005266970.1 | XP_005267027.1 | 13 | 6868 | |||
XM_005266971.1 | XP_005267028.1 | 13 | 6831 | |||
XM_005266972.4 | XP_005267029.1 | 11 | 6971 | |||
XM_006715475.3 | XP_006715538.1 | 14 | 7044 | |||
XM_011535809.2 | XP_011534111.1 | 14 | 7939 | |||
XM_011535810.2 | XP_011534112.1 | 14 | 6793 | |||
XM_011535811.1 | XP_011534113.1 | 13 | 6734 | |||
XM_017010846.1 | XP_016866335.1 | 15 | 6972 | |||
XM_017010847.1 | XP_016866336.1 | 12 | 6983 | |||
GenBank mRNA | AL133101.1 | - | 1 | 2409 | ||
BC045657.1 | - | 1 | 3934 |