Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ID4 SIRNA Assays ›
          Gene Symbol:
          ID4
          Gene Name:
          inhibitor of DNA binding 4, HLH protein
          Gene Aliases:
          IDB4, bHLHb27
          Chromosome Location:
          Chr.6: 19837370 - 19842200 on Build GRCh38
          UniGene ID:
          Hs.519601
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n280974

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ID4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001546.3 NP_001537.1 3 3774
          GenBank mRNA AJ420553.1 - 1 2026
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB532145.1 - 1 334 Chr. 6 - 19841766 - 19842193 [+] on Build GRCh38
          GenBankmRNA DB552982.1 - 1 364 Chr. 6 - 19841735 - 19842193 [+] on Build GRCh38
          GenBankmRNA DB541000.1 - 1 363 Chr. 6 - 19841737 - 19842193 [+] on Build GRCh38
          GenBankmRNA DB566997.1 - 1 371 Chr. 6 - 19841728 - 19842193 [+] on Build GRCh38
          GenBankmRNA DB552262.1 - 1 367 Chr. 6 - 19841732 - 19842193 [+] on Build GRCh38
          GenBankmRNA DB546610.1 - 1 328 Chr. 6 - 19841772 - 19842193 [+] on Build GRCh38
          GenBankmRNA CA942347.1 - 1 590 Chr. 6 - 19841512 - 19842121 [+] on Build GRCh38
          GenBankmRNA DB551779.1 - 1 373 Chr. 6 - 19841726 - 19842193 [+] on Build GRCh38
          GenBankmRNA D62553.1 - 1 298 Chr. 6 - 19841800 - 19842193 [+] on Build GRCh38
          GenBankmRNA BP391503.1 - 1 296 Chr. 6 - 19841803 - 19842191 [+] on Build GRCh38
          GenBankmRNA DB547205.1 - 1 330 Chr. 6 - 19841769 - 19842193 [+] on Build GRCh38
          GenBankmRNA DB527283.1 - 1 375 Chr. 6 - 19841724 - 19842193 [+] on Build GRCh38
          GenBankmRNA DB552851.1 - 1 358 Chr. 6 - 19841740 - 19842193 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          neuroblast proliferation
          circadian rhythm
          positive regulation of cell proliferation
          hippocampus development
          cerebral cortex neuron differentiation
          central nervous system myelination
          cellular protein localization
          fat cell differentiation
          negative regulation of fat cell differentiation
          negative regulation of neuron differentiation
          positive regulation of osteoblast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of astrocyte differentiation
          negative regulation of oligodendrocyte differentiation
          prostate gland epithelium morphogenesis
          prostate gland stromal morphogenesis
          seminal vesicle morphogenesis
          RNA polymerase II transcription factor binding
          transcription corepressor activity
          protein binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-brsnx:80/100.66.78.229:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0