Search
Search

LOC283575| Gene Symbol: LOC283575 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110554.1 | 4 | 2106 | |||
| GenBank mRNA | AK092322.1 | - | 4 | 2091 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000500215.3 | - | 4 | 2091 | Chr. 14 - 77041064 - 77069503 [+] on Build GRCh38 | |