Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGD5-AS1 SIRNA Assays ›
          Gene Symbol:
          FGD5-AS1
          Gene Name:
          FGD5 antisense RNA 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 14942779 - 14948441 on Build GRCh38
          UniGene ID:
          Hs.517821
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n283704

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FGD5-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_046251.1 2 3181
          NR_046252.1 2 3171
          NR_046253.1 2 3148
          NR_046254.1 2 3144
          NR_046255.1 2 2907
          GenBank mRNA BC039246.1 - 2 3131
          BC042881.1 - 1 301
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BX348625.2 - 1 441 Chr. 3 - 14942933 - 14943839 [-] on Build GRCh38
          GenBankmRNA BM781774.1 - 1 101 Chr. 3 - 14942897 - 14943503 [-] on Build GRCh38
          GenBankmRNA R70976.1 - 18 Chr. - - [-] on Build
          GenBankmRNA CD678137.1 - 1 294 Chr. 3 - 14943062 - 14943696 [-] on Build GRCh38
          GenBankmRNA BM723448.1 - 1 254 Chr. 3 - 14942990 - 14943656 [-] on Build GRCh38
          GenBankmRNA HY108212.1 - 1 46 Chr. 3 - 14942980 - 14943447 [-] on Build GRCh38
          GenBankmRNA BX348624.1 - 1 432 Chr. 3 - 14942932 - 14943799 [-] on Build GRCh38
          GenBankmRNA AA405589.1 - 1 29 Chr. 3 - 14943012 - 14943431 [-] on Build GRCh38
          GenBankmRNA BM555403.1 - 1 221 Chr. 3 - 14942784 - 14943593 [-] on Build GRCh38
          GenBankmRNA CB998796.1 - 1 276 Chr. 3 - 14942940 - 14943676 [-] on Build GRCh38
          GenBankmRNA BG026664.1 - 386 Chr. - - [-] on Build
          GenBankmRNA CN370579.1 - 1 255 Chr. 3 - 14943135 - 14943657 [-] on Build GRCh38
          GenBankmRNA BM721227.1 - 1 23 Chr. 3 - 14942779 - 14943425 [-] on Build GRCh38
          GenBankmRNA AA383814.1 - 1 25 Chr. 3 - 14943136 - 14943419 [-] on Build GRCh38
          GenBankmRNA BG029393.1 - 1 171 Chr. 3 - 14942788 - 14943573 [-] on Build GRCh38
          GenBankmRNA BG111940.1 - 1 28 Chr. 3 - 14942781 - 14943419 [-] on Build GRCh38
          GenBankmRNA BM762106.1 - 1 101 Chr. 3 - 14942856 - 14943503 [-] on Build GRCh38
          GenBankmRNA CR738841.1 - 1 65 Chr. 3 - 14942958 - 14943467 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zkdp8:80/100.66.75.244:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0