Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RAB2A SIRNA Assays ›
          Gene Symbol:
          RAB2A
          Gene Name:
          RAB2A, member RAS oncogene family
          Gene Aliases:
          LHX, RAB2
          Chromosome Location:
          Chr.8: 60516910 - 60623644 on Build GRCh38
          UniGene ID:
          Hs.369017
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n284651

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RAB2A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001242644.1 NP_001229573.1 7 3049
          NM_002865.2 NP_002856.1 8 3121
          GenBank mRNA AL137321.1 - 1 1857
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM817586.1 - 1 276 Chr. 8 - 60622678 - 60623098 [+] on Build GRCh38
          GenBankmRNA DB548715.1 - 1 166 Chr. 8 - 60622788 - 60623239 [+] on Build GRCh38
          GenBankmRNA DB546419.1 - 1 252 Chr. 8 - 60622716 - 60623094 [+] on Build GRCh38
          GenBankmRNA CD641108.1 - 281 Chr. - - [-] on Build
          GenBankmRNA BG483092.1 - 1 409 Chr. 8 - 60622546 - 60623300 [+] on Build GRCh38
          GenBankmRNA DB541955.1 - 1 331 Chr. 8 - 60622623 - 60623090 [+] on Build GRCh38
          GenBankmRNA DB534626.1 - 1 287 Chr. 8 - 60622668 - 60623100 [+] on Build GRCh38
          GenBankmRNA DB567029.1 - 1 328 Chr. 8 - 60622626 - 60623097 [+] on Build GRCh38
          GenBankmRNA BM723508.1 - 1 245 Chr. 8 - 60622709 - 60623112 [+] on Build GRCh38
          GenBankmRNA DB524779.1 - 1 342 Chr. 8 - 60622612 - 60623090 [+] on Build GRCh38
          GenBankmRNA DB538670.1 - 1 293 Chr. 8 - 60622659 - 60623100 [+] on Build GRCh38
          GenBankmRNA CB155677.1 - 1 458 Chr. 8 - 60622496 - 60623022 [+] on Build GRCh38
          GenBankmRNA DB541778.1 - 1 184 Chr. 8 - 60622770 - 60623239 [+] on Build GRCh38
          GenBankmRNA DB561003.1 - 1 255 Chr. 8 - 60622699 - 60623094 [+] on Build GRCh38
          GenBankmRNA DB534106.1 - 1 275 Chr. 8 - 60622678 - 60623100 [+] on Build GRCh38
          GenBankmRNA DA261497.1 - 1 279 Chr. 8 - 60622676 - 60623260 [+] on Build GRCh38
          GenBankmRNA BG181098.1 - 1 236 Chr. 8 - 60622717 - 60623367 [+] on Build GRCh38
          GenBankmRNA DB522715.1 - 1 313 Chr. 8 - 60622641 - 60623094 [+] on Build GRCh38
          GenBankmRNA BM505056.1 - 1 397 Chr. 8 - 60622557 - 60623066 [+] on Build GRCh38
          GenBankmRNA DB543747.1 - 1 309 Chr. 8 - 60622646 - 60623090 [+] on Build GRCh38
          GenBankmRNA DB557935.1 - 1 296 Chr. 8 - 60622658 - 60623094 [+] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ER to Golgi vesicle-mediated transport
          Golgi organization
          small GTPase mediated signal transduction
          protein transport
          GTPase activity
          protein binding
          GTP binding
          GDP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0