Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SLC35F5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005263799.3 | XP_005263856.1 | 16 | 8498 | ||
XM_011511921.2 | XP_011510223.1 | 17 | 7949 | |||
XM_011511922.2 | XP_011510224.1 | 17 | 8166 | |||
XM_011511923.2 | XP_011510225.1 | 16 | 7851 | |||
XM_017005028.1 | XP_016860517.1 | 15 | 7812 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK093903.1 | - | 1 | 1149 | Chr. 2 - 113706782 - 113710239 [-] on Build GRCh38 |