Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MZF1 SIRNA Assays ›
          Gene Symbol:
          MZF1
          Gene Name:
          myeloid zinc finger 1
          Gene Aliases:
          MZF-1, MZF1B, ZFP98, ZNF42, ZSCAN6
          Chromosome Location:
          Chr.19: 58561917 - 58573851 on Build GRCh38
          UniGene ID:
          Hs.399810
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n290344

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MZF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA M58297.1 - 1 140
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BE327149.1 - 1 408 Chr. 19 - 58564363 - 58564887 [-] on Build GRCh38
          GenBankmRNA BJ990573.1 - 1 217 Chr. 19 - 58564099 - 58564687 [-] on Build GRCh38
          GenBankmRNA CN276853.1 - 1 261 Chr. 19 - 58564229 - 58564742 [-] on Build GRCh38
          GenBankmRNA BX336058.2 - 1 415 Chr. 19 - 58564021 - 58564896 [-] on Build GRCh38
          GenBankmRNA BX441810.2 - 1 425 Chr. 19 - 58564015 - 58564906 [-] on Build GRCh38
          GenBankmRNA AI820605.1 - 1 395 Chr. 19 - 58564353 - 58564876 [-] on Build GRCh38
          GenBankmRNA BF448206.1 - 1 411 Chr. 19 - 58564371 - 58564890 [-] on Build GRCh38
          GenBankmRNA AI733248.1 - 1 406 Chr. 19 - 58564267 - 58564887 [-] on Build GRCh38
          GenBankmRNA CN276854.1 - 1 186 Chr. 19 - 58564178 - 58564667 [-] on Build GRCh38
          GenBankmRNA AI300989.1 - 1 408 Chr. 19 - 58564448 - 58564887 [-] on Build GRCh38
          GenBankmRNA AI457383.1 - 1 426 Chr. 19 - 58564342 - 58564887 [-] on Build GRCh38
          GenBankmRNA AW367884.1 - 1 175 Chr. 19 - 58564151 - 58564651 [-] on Build GRCh38
          GenBankmRNA BE222264.1 - 1 408 Chr. 19 - 58564282 - 58564886 [-] on Build GRCh38
          GenBankmRNA AW892671.1 - 117 Chr. - - [-] on Build
          GenBankmRNA BU070843.1 - 1 379 Chr. 19 - 58564398 - 58564853 [-] on Build GRCh38
          GenBankmRNA AI459062.1 - 1 407 Chr. 19 - 58564320 - 58564887 [-] on Build GRCh38
          GenBankmRNA BE467998.1 - 1 411 Chr. 19 - 58564371 - 58564890 [-] on Build GRCh38
          GenBankmRNA AA987906.1 - 1 399 Chr. 19 - 58564328 - 58564876 [-] on Build GRCh38
          GenBankmRNA AA019979.1 - 1 118 Chr. 19 - 58564323 - 58564597 [-] on Build GRCh38
          GenBankmRNA AI871765.1 - 1 396 Chr. 19 - 58564379 - 58564866 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-zkdp8:80/100.66.75.244:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0