Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MYO15B SIRNA Assays ›
          Gene Symbol:
          MYO15B
          Gene Name:
          myosin XVB
          Gene Aliases:
          MYO15BP
          Chromosome Location:
          Chr.17: 75587545 - 75626850 on Build GRCh38
          UniGene ID:
          Hs.390817
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n296589

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MYO15B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001309242.1 NP_001296171.1 22 4317
          XM_017025120.1 XP_016880609.1 24 4024
          XM_017025121.1 XP_016880610.1 24 4009
          XM_017025122.1 XP_016880611.1 24 3988
          XM_017025123.1 XP_016880612.1 24 4026
          XM_017025124.1 XP_016880613.1 23 3898
          XM_017025125.1 XP_016880614.1 22 3747
          XM_017025126.1 XP_016880615.1 19 3489
          XM_017025127.1 XP_016880616.1 22 3332
          XM_017025128.1 XP_016880617.1 23 3078
          XM_017025129.1 XP_016880618.1 22 2310
          XM_017025130.1 XP_016880619.1 21 2266
          XM_017025131.1 XP_016880620.1 20 2705
          XM_017025132.1 XP_016880621.1 19 2646
          XM_017025133.1 XP_016880622.1 17 2171
          XM_017025134.1 XP_016880623.1 18 2532
          XM_017025135.1 XP_016880624.1 12 1188
          XM_017025136.1 XP_016880625.1 12 1537
          XM_017025137.1 XP_016880626.1 9 1082
          XM_017025138.1 XP_016880627.1 4 578
          XM_017025139.1 XP_016880628.1 5 379
          XM_017025140.1 XP_016880629.1 2 192
          XM_017025141.1 XP_016880630.1 24 4290
          XM_017025142.1 XP_016880631.1 24 4290
          XM_017025144.1 XP_016880633.1 17 3899
          GenBank mRNA AB058686.1 - 12 2424
          AF418286.1 - 20 4550
          AF418288.1 - 21 4334
          AF418289.1 - 17 3899
          AF418290.1 - 17 4854
          AF418291.1 - 17 3900
          AK096869.1 - 1 25
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BE767401.1 - 2 149 Chr. 17 - 75610193 - 75612864 [+] on Build GRCh38
          GenBankmRNA HY166912.1 - 2 101 Chr. 17 - 75610676 - 75613077 [+] on Build GRCh38
          GenBankmRNA BM906425.1 - 125 Chr. - - [-] on Build
          GenBankmRNA BX401175.2 - 1 46 Chr. 17 - 75610899 - 75614270 [+] on Build GRCh38
          GenBankmRNA BM737684.1 - 2 70 Chr. 17 - 75610707 - 75613190 [+] on Build GRCh38
          GenBankmRNA DA890157.1 - 1 25 Chr. 17 - 75610922 - 75613211 [+] on Build GRCh38
          GenBankmRNA BM737740.1 - 2 67 Chr. 17 - 75610710 - 75613000 [+] on Build GRCh38
          GenBankmRNA BM738296.1 - 2 78 Chr. 17 - 75610699 - 75613107 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          actin binding motor protein

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0