Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGF10 SIRNA Assays ›
Gene Symbol:
FGF10
Gene Name:
fibroblast growth factor 10
Gene Aliases:
-
Chromosome Location:
Chr.5: 44301655 - 44389780 on Build GRCh38
UniGene ID:
Hs.248049
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n301579
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGF10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_005248264.3
XP_005248321.1
4
2890
GenBank mRNA
AK023508.1
-
1
343
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
establishment of mitotic spindle orientation
MAPK cascade
activation of MAPK activity
angiogenesis
metanephros development
organ induction
mesonephros development
blood vessel remodeling
metanephros morphogenesis
determination of left/right symmetry
salivary gland development
negative regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
regulation of smoothened signaling pathway
embryonic pattern specification
epithelial cell migration
positive regulation of epithelial cell migration
positive regulation of keratinocyte proliferation
regulation of phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
pituitary gland development
embryonic genitalia morphogenesis
thyroid gland development
otic vesicle formation
positive regulation of vascular endothelial growth factor receptor signaling pathway
pancreas development
hair follicle morphogenesis
embryonic camera-type eye development
actin cytoskeleton reorganization
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
response to estradiol
response to lipopolysaccharide
positive regulation of ATPase activity
lacrimal gland development
regulation of activin receptor signaling pathway
protein localization to cell surface
somatic stem cell population maintenance
phosphatidylinositol-3-phosphate biosynthetic process
wound healing
tissue regeneration
odontogenesis of dentin-containing tooth
muscle cell fate commitment
positive regulation of MAPK cascade
positive regulation of GTPase activity
keratinocyte proliferation
negative regulation of cell differentiation
positive regulation of DNA repair
positive regulation of DNA replication
positive regulation of Notch signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of Ras protein signal transduction
phosphatidylinositol phosphorylation
regulation of saliva secretion
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
spleen development
thymus development
embryonic digestive tract morphogenesis
branching morphogenesis of an epithelial tube
female genitalia morphogenesis
male genitalia morphogenesis
positive regulation of lymphocyte proliferation
epithelial cell proliferation
urothelial cell proliferation
positive regulation of urothelial cell proliferation
positive regulation of epithelial cell proliferation
positive regulation of peptidyl-tyrosine phosphorylation
white fat cell differentiation
positive chemotaxis
induction of positive chemotaxis
smooth muscle cell differentiation
positive regulation of keratinocyte migration
radial glial cell differentiation
positive regulation of epithelial cell proliferation involved in wound healing
limb bud formation
lung epithelium development
lung saccule development
bronchiole morphogenesis
bud outgrowth involved in lung branching
bud elongation involved in lung branching
mesenchymal-epithelial cell signaling involved in lung development
Type II pneumocyte differentiation
prostatic bud formation
fibroblast growth factor receptor signaling pathway involved in mammary gland specification
mammary gland bud formation
submandibular salivary gland formation
epithelial cell proliferation involved in salivary gland morphogenesis
regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
branch elongation involved in salivary gland morphogenesis
semicircular canal fusion
mesenchymal cell differentiation involved in lung development
secretion by lung epithelial cell involved in lung growth
lung proximal/distal axis specification
tear secretion
positive regulation of white fat cell proliferation
ERK1 and ERK2 cascade
positive regulation of ERK1 and ERK2 cascade
Harderian gland development
negative regulation of cell cycle arrest
positive regulation of hair follicle cell proliferation
positive regulation of canonical Wnt signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
fibroblast growth factor receptor binding
type 2 fibroblast growth factor receptor binding
protein binding
growth factor activity
heparin binding
1-phosphatidylinositol-3-kinase activity
chemoattractant activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0