Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LINC00641 SIRNA Assays ›
          Gene Symbol:
          LINC00641
          Gene Name:
          long intergenic non-protein coding RNA 641
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 21200079 - 21206900 on Build GRCh38
          UniGene ID:
          Hs.525210
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n310859

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LINC00641
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_038970.1 1 1865
          GenBank mRNA AK092333.1 - 1 717
          BC035145.1 - 1 1868
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DB220976.1 - 1 276 Chr. 14 - 21203369 - 21203914 [-] on Build GRCh38
          GenBankmRNA DA101949.1 - 1 24 Chr. 14 - 21203106 - 21203662 [-] on Build GRCh38
          GenBankmRNA DA758949.1 - 1 412 Chr. 14 - 21203506 - 21204050 [-] on Build GRCh38
          GenBankmRNA DA212205.1 - 1 284 Chr. 14 - 21203379 - 21203922 [-] on Build GRCh38
          GenBankmRNA DA405800.1 - 1 190 Chr. 14 - 21203267 - 21203828 [-] on Build GRCh38
          GenBankmRNA DA315470.1 - 1 241 Chr. 14 - 21203339 - 21203879 [-] on Build GRCh38
          GenBankmRNA DA118558.1 - 1 136 Chr. 14 - 21203244 - 21203774 [-] on Build GRCh38
          GenBankmRNA DA289628.1 - 1 41 Chr. 14 - 21203100 - 21203679 [-] on Build GRCh38
          GenBankmRNA DA118168.1 - 1 343 Chr. 14 - 21203418 - 21203979 [-] on Build GRCh38
          GenBankmRNA DA243125.1 - 1 137 Chr. 14 - 21203183 - 21203775 [-] on Build GRCh38
          GenBankmRNA DA326033.1 - 1 345 Chr. 14 - 21203467 - 21203981 [-] on Build GRCh38
          GenBankmRNA DA247375.1 - 1 132 Chr. 14 - 21203194 - 21203770 [-] on Build GRCh38
          GenBankmRNA DA530605.1 - 1 189 Chr. 14 - 21203209 - 21203827 [-] on Build GRCh38
          GenBankmRNA DA219413.1 - 1 191 Chr. 14 - 21203369 - 21203828 [-] on Build GRCh38
          GenBankmRNA DA328986.1 - 1 243 Chr. 14 - 21203330 - 21203880 [-] on Build GRCh38
          GenBankmRNA DA099752.1 - 1 421 Chr. 14 - 21203483 - 21204058 [-] on Build GRCh38
          GenBankmRNA DA236739.1 - 1 411 Chr. 14 - 21203503 - 21204049 [-] on Build GRCh38
          GenBankmRNA DA247398.1 - 1 132 Chr. 14 - 21203199 - 21203770 [-] on Build GRCh38
          GenBankmRNA DA391104.1 - 1 140 Chr. 14 - 21203191 - 21203778 [-] on Build GRCh38
          GenBankmRNA DA117140.1 - 1 144 Chr. 14 - 21203233 - 21203782 [-] on Build GRCh38
          GenBankmRNA DA338264.1 - 1 266 Chr. 14 - 21203310 - 21203904 [-] on Build GRCh38
          GenBankmRNA BX504977.1 - 1 63 Chr. 14 - 21203252 - 21203701 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0