Search
Search

RBPMS| Gene Symbol: RBPMS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012984.1 | XP_016868473.1 | 6 | 1198 | ||
| GenBank mRNA | AK057533.1 | - | 1 | 1074 | ||
| AK304858.1 | - | 7 | 1021 | |||
| AK310533.1 | - | 6 | 631 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX952240.1 | - | 1 | 397 | Chr. 8 - 30544311 - 30544780 [+] on Build GRCh38 | |