Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011537552.2 | XP_011535854.1 | 3 | 5784 | ||
GenBank mRNA | AK095279.1 | - | 1 | 1389 |