Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARPP21 SIRNA Assays ›
Gene Symbol:
ARPP21
LOC100130503
Gene Name:
cAMP regulated phosphoprotein 21
uncharacterized LOC100130503
Gene Aliases:
ARPP-21, R3HDM3, RCS, TARPP
-
Chromosome Location:
Chr.3: 35638981 - 35798933 on Build GRCh38
Chromosome Location:
-
UniGene ID:
Hs.475902
Hs.475902
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
n329348
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARPP21
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001267616.1
NP_001254545.1
1
732
XM_011533299.2
XP_011531601.1
1
132
XM_011533303.2
XP_011531605.1
1
132
XM_017005576.1
XP_016861065.1
1
132
XM_017005579.1
XP_016861068.1
1
132
XM_017005580.1
XP_016861069.1
1
132
XM_017005582.1
XP_016861071.1
1
132
XM_017005584.1
XP_016861073.1
1
132
XM_017005585.1
XP_016861074.1
1
132
XM_017005588.1
XP_016861077.1
1
132
XM_017005591.1
XP_016861080.1
1
132
XM_017005592.1
XP_016861081.1
1
132
XM_017005595.1
XP_016861084.1
1
132
XM_017005596.1
XP_016861085.1
1
132
XM_017005597.1
XP_016861086.1
1
132
XM_017005598.1
XP_016861087.1
1
132
XM_017005599.1
XP_016861088.1
1
132
XM_017005602.1
XP_016861091.1
1
132
XM_017005605.1
XP_016861094.1
1
132
XM_017005607.1
XP_016861096.1
1
132
XM_017005608.1
XP_016861097.1
1
132
XM_017005609.1
XP_016861098.1
1
132
XM_017005610.1
XP_016861099.1
1
132
XM_017005611.1
XP_016861100.1
1
132
XM_017005612.1
XP_016861101.1
1
132
GenBank mRNA
BC041385.1
-
1
320
Gene Symbol: LOC100130503
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
AK057559.1
-
1
202
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
cellular response to heat
molecular_function
nucleic acid binding
calmodulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0