Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RUNX1T1 SIRNA Assays ›
Gene Symbol:
RUNX1T1
Gene Name:
RUNX1 translocation partner 1
Gene Aliases:
AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
Chromosome Location:
Chr.8: 91954967 - 92103365 on Build GRCh38
UniGene ID:
Hs.368431
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n335478
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
5,354.00
Your Price
Online offer:
5354.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RUNX1T1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198625.1
NP_001185554.1
15
7546
NM_001198626.1
NP_001185555.1
13
7404
NM_001198627.1
NP_001185556.1
14
7464
NM_001198628.1
NP_001185557.1
12
7061
NM_001198629.1
NP_001185558.1
12
7187
NM_001198630.1
NP_001185559.1
13
7247
NM_001198631.1
NP_001185560.1
12
7260
NM_001198632.1
NP_001185561.1
12
7178
NM_001198633.1
NP_001185562.1
11
7149
NM_001198634.1
NP_001185563.1
11
7087
NM_001198679.1
NP_001185608.1
12
7231
NM_004349.3
NP_004340.1
11
7197
NM_175634.2
NP_783552.1
12
7322
NM_175635.2
NP_783553.1
11
7014
NM_175636.2
NP_783554.1
10
7096
XM_006716676.3
XP_006716739.1
13
7131
XM_011517351.2
XP_011515653.1
13
7176
XM_011517352.2
XP_011515654.1
13
7416
XM_011517353.2
XP_011515655.1
12
7124
XM_017013930.1
XP_016869419.1
12
7432
XM_017013931.1
XP_016869420.1
11
6954
XM_017013932.1
XP_016869421.1
13
7319
XM_017013933.1
XP_016869422.1
14
7383
XM_017013934.1
XP_016869423.1
14
7491
XM_017013935.1
XP_016869424.1
13
7296
XM_017013936.1
XP_016869425.1
12
7390
XM_017013937.1
XP_016869426.1
13
7216
XM_017013939.1
XP_016869428.1
7
6847
XM_017013940.1
XP_016869429.1
8
8076
XM_017013941.1
XP_016869430.1
7
7866
GenBank mRNA
AL049240.1
-
1
538
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
generation of precursor metabolites and energy
transcription, DNA-templated
negative regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription corepressor activity
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0