Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ZNF827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306215.1 | NP_001293144.1 | 1 | 198 | ||
NM_178835.4 | NP_849157.2 | 1 | 198 | |||
XM_011531634.2 | XP_011529936.1 | 1 | 245 | |||
XM_017007768.1 | XP_016863257.1 | 1 | 246 | |||
XM_017007770.1 | XP_016863259.1 | 1 | 246 | |||
XM_017007771.1 | XP_016863260.1 | 1 | 244 | |||
XM_017007772.1 | XP_016863261.1 | 1 | 244 | |||
XM_017007773.1 | XP_016863262.1 | 1 | 244 | |||
XM_017007774.1 | XP_016863263.1 | 1 | 244 | |||
XM_017007775.1 | XP_016863264.1 | 1 | 244 | |||
XM_017007776.1 | XP_016863265.1 | 1 | 246 | |||
GenBank mRNA | BC033879.1 | - | 1 | 57 | ||
BM557137.1 | - | 1 | 45 | |||
BX112429.1 | - | 1 | 198 |