Search
Search

COA1| Gene Symbol: COA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_005249807.4 | XP_005249864.1 | 7 | 9720 | ||
| XM_006715752.2 | XP_006715815.1 | 8 | 9688 | |||
| XM_017012414.1 | XP_016867903.1 | 7 | 9647 | |||
| XM_017012415.1 | XP_016867904.1 | 8 | 9749 | |||
| XM_017012417.1 | XP_016867906.1 | 8 | 8151 | |||
| XM_017012418.1 | XP_016867907.1 | 7 | 8021 | |||
| XM_017012421.1 | XP_016867910.1 | 8 | 9871 | |||
| XM_017012422.1 | XP_016867911.1 | 8 | 9760 | |||
| XM_017012424.1 | XP_016867913.1 | 7 | 9663 | |||
| XM_017012430.1 | XP_016867919.1 | 8 | 8123 | |||
| GenBank mRNA | BX537586.1 | - | 1 | 3184 | ||