Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
Unity Lab Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GPR157 SIRNA Assays ›
Gene Symbol:
GPR157
Gene Name:
G protein-coupled receptor 157
Gene Aliases:
-
Chromosome Location:
Chr.1: 9103390 - 9129170 on Build GRCh38
UniGene ID:
Hs.632367
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n338610
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GPR157
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_005263496.4
XP_005263553.1
4
2001
XM_005263497.4
XP_005263554.1
3
1845
GenBank mRNA
AK026883.1
-
1
930
Back To Top
More Information
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0