Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RAB3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317915.1 | NP_001304844.1 | 5 | 7597 | ||
NM_138453.3 | NP_612462.1 | 5 | 7559 | |||
GenBank mRNA | AB073648.1 | - | 1 | 729 | ||
AB074188.1 | - | 1 | 554 | |||
AK055409.1 | - | 1 | 760 | |||
AK094685.1 | - | 1 | 1606 |