Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 企業服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTOV1-AS2 SIRNA Assays ›
          Gene Symbol:
          PTOV1-AS2
          Gene Name:
          PTOV1 antisense RNA 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 49856465 - 49859289 on Build GRCh38
          UniGene ID:
          Hs.710295
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n355027

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTOV1-AS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AL050131.1 - 1 433
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CX755546.1 - 6 649 Chr. 19 - 49857742 - 49860738 [-] on Build GRCh38
          GenBankmRNA AL520297.3 - 6 565 Chr. 19 - 49854975 - 49860708 [-] on Build GRCh38
          GenBankmRNA BM972673.1 - 6 606 Chr. 19 - 49857912 - 49860742 [-] on Build GRCh38
          GenBankmRNA DR156022.1 - 6 611 Chr. 19 - 49857753 - 49860742 [-] on Build GRCh38
          GenBankmRNA CX163151.1 - 6 658 Chr. 19 - 49857741 - 49860740 [-] on Build GRCh38
          GenBankmRNA CX760209.1 - 6 609 Chr. 19 - 49857682 - 49860742 [-] on Build GRCh38
          GenBankmRNA CR999807.1 - 1 467 Chr. 19 - 49857736 - 49858417 [-] on Build GRCh38
          GenBankmRNA BI522052.1 - 6 532 Chr. 19 - 49855013 - 49860668 [-] on Build GRCh38
          GenBankmRNA BM682306.1 - 6 496 Chr. 19 - 49857907 - 49860742 [-] on Build GRCh38
          GenBankmRNA BM808224.1 - 470 Chr. - - [-] on Build
          GenBankmRNA CR986487.1 - 1 315 Chr. 19 - 49857570 - 49858262 [-] on Build GRCh38
          GenBankmRNA BM981376.1 - 6 651 Chr. 19 - 49857914 - 49860740 [-] on Build GRCh38
          GenBankmRNA BC028302.1 - 6 601 Chr. 19 - 49851387 - 49860743 [-] on Build GRCh38
          GenBankmRNA CB529565.1 - 6 651 Chr. 19 - 49857104 - 49860742 [-] on Build GRCh38
          GenBankmRNA DA995751.1 - 1 184 Chr. 19 - 49857562 - 49858134 [-] on Build GRCh38
          GenBankmRNA BQ439673.1 - 625 Chr. - - [-] on Build
          GenBankmRNA DR763683.1 - 6 585 Chr. 19 - 49857077 - 49860739 [-] on Build GRCh38
          GenBankmRNA CX872788.1 - 6 612 Chr. 19 - 49857116 - 49860742 [-] on Build GRCh38
          GenBankmRNA DN989559.1 - 1 260 Chr. 19 - 49857677 - 49858203 [-] on Build GRCh38
          GenBankmRNA BU619943.1 - 6 534 Chr. 19 - 49857712 - 49860742 [-] on Build GRCh38
          GenBankmRNA BX102458.1 - 6 546 Chr. 19 - 49857054 - 49860701 [-] on Build GRCh38
          GenBankmRNA BG820502.1 - 1 337 Chr. 19 - 49857838 - 49858285 [-] on Build GRCh38
          GenBankmRNA BF966827.1 - 6 550 Chr. 19 - 49857071 - 49860692 [-] on Build GRCh38
          GenBankmRNA BM677857.1 - 6 604 Chr. 19 - 49857709 - 49860742 [-] on Build GRCh38
          GenBankmRNA BQ183847.1 - 6 603 Chr. 19 - 49857706 - 49860740 [-] on Build GRCh38
          GenBankmRNA BU625568.1 - 6 604 Chr. 19 - 49857902 - 49860742 [-] on Build GRCh38
          GenBankmRNA BU731573.1 - 6 602 Chr. 19 - 49857913 - 49860740 [-] on Build GRCh38
          GenBankmRNA BF570480.1 - 6 547 Chr. 19 - 49857104 - 49860673 [-] on Build GRCh38
          GenBankmRNA AW245962.1 - 6 594 Chr. 19 - 49857923 - 49860741 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0