Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MAGI2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038343.2 | 4 | 2598 | |||
GenBank mRNA | AK021837.1 | - | 1 | 62 | ||
AL833456.1 | - | 2 | 2323 | |||
BX641113.1 | - | 1 | 1490 | |||
CR749529.1 | - | 2 | 2651 |