Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006712499.3 | XP_006712562.1 | 26 | 3770 | ||
XM_006712500.3 | XP_006712563.1 | 26 | 3767 | |||
XM_006712501.3 | XP_006712564.1 | 25 | 3752 | |||
XM_006712502.3 | XP_006712565.1 | 25 | 3737 | |||
XM_006712504.3 | XP_006712567.1 | 26 | 3653 | |||
XM_006712506.3 | XP_006712569.1 | 25 | 3620 | |||
XM_011511122.2 | XP_011509424.1 | 25 | 3720 | |||
XM_011511125.2 | XP_011509427.1 | 23 | 3652 | |||
XM_011511127.2 | XP_011509429.1 | 14 | 2455 | |||
XM_017003993.1 | XP_016859482.1 | 26 | 3494 | |||
XM_017003994.1 | XP_016859483.1 | 25 | 3635 | |||
XM_017003995.1 | XP_016859484.1 | 25 | 3617 | |||
GenBank mRNA | BC005082.1 | - | 1 | 1672 |