Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FAM78B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320302.1 | NP_001307231.1 | 2 | 3918 | ||
NR_135199.1 | 3 | 4850 | ||||
XM_017000417.1 | XP_016855906.1 | 3 | 9056 | |||
XM_017000418.1 | XP_016855907.1 | 3 | 4398 | |||
GenBank mRNA | AB593134.1 | - | 2 | 3921 | ||
AK055347.1 | - | 1 | 1954 |