Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERO1A SIRNA Assays ›
          Gene Symbol:
          ERO1A
          Gene Name:
          endoplasmic reticulum oxidoreductase 1 alpha
          Gene Aliases:
          ERO1-L, ERO1-L-alpha, ERO1-alpha, ERO1L, ERO1LA, Ero1alpha
          Chromosome Location:
          Chr.14: 52639915 - 52695931 on Build GRCh38
          UniGene ID:
          Hs.525339
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n363155

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ERO1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014584.2 NP_055399.1 16 4342
          XM_011536684.2 XP_011534986.1 15 4149
          GenBank mRNA AK024224.1 - 1 727
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CD172532.1 - 336 Chr. - - [-] on Build
          GenBankmRNA BM843136.1 - 1 110 Chr. 14 - 52640709 - 52641194 [-] on Build GRCh38
          GenBankmRNA H66960.1 - 1 36 Chr. 14 - 52640704 - 52641120 [-] on Build GRCh38
          GenBankmRNA BM844697.1 - 1 109 Chr. 14 - 52640657 - 52641191 [-] on Build GRCh38
          GenBankmRNA BM851360.1 - 1 110 Chr. 14 - 52640633 - 52641194 [-] on Build GRCh38
          GenBankmRNA BU674223.1 - 1 234 Chr. 14 - 52640640 - 52641317 [-] on Build GRCh38
          GenBankmRNA BM761748.1 - 1 110 Chr. 14 - 52640621 - 52641194 [-] on Build GRCh38
          GenBankmRNA BM845971.1 - 1 110 Chr. 14 - 52640777 - 52641194 [-] on Build GRCh38
          GenBankmRNA DA728236.1 - 1 240 Chr. 14 - 52640520 - 52641317 [-] on Build GRCh38
          GenBankmRNA BU607701.1 - 1 241 Chr. 14 - 52640599 - 52641319 [-] on Build GRCh38
          GenBankmRNA BM761643.1 - 1 110 Chr. 14 - 52640527 - 52641194 [-] on Build GRCh38
          GenBankmRNA AW965455.1 - 1 205 Chr. 14 - 52640695 - 52641289 [-] on Build GRCh38
          GenBankmRNA BU685489.1 - 1 240 Chr. 14 - 52640615 - 52641319 [-] on Build GRCh38
          GenBankmRNA BM847261.1 - 1 110 Chr. 14 - 52640623 - 52641194 [-] on Build GRCh38
          GenBankmRNA BQ717224.1 - 1 315 Chr. 14 - 52640562 - 52641399 [-] on Build GRCh38
          GenBankmRNA CN302088.1 - 1 224 Chr. 14 - 52640738 - 52641301 [-] on Build GRCh38
          GenBankmRNA CA773350.1 - 1 154 Chr. 14 - 52640643 - 52641238 [-] on Build GRCh38
          GenBankmRNA BM761670.1 - 1 110 Chr. 14 - 52640587 - 52641194 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to reactive oxygen species
          protein folding
          cellular protein modification process
          response to temperature stimulus
          organ senescence
          4-hydroxyproline metabolic process
          protein maturation by protein folding
          extracellular matrix organization
          endoplasmic reticulum unfolded protein response
          response to endoplasmic reticulum stress
          cell redox homeostasis
          brown fat cell differentiation
          chaperone mediated protein folding requiring cofactor
          release of sequestered calcium ion into cytosol
          oxidation-reduction process
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          cellular response to hypoxia
          protein disulfide isomerase activity
          protein binding
          protein disulfide oxidoreductase activity
          disulfide oxidoreductase activity
          oxidoreductase activity
          oxidoreductase activity, acting on a sulfur group of donors, disulfide as acceptor

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-dvg58:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0