Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TFDP2 SIRNA Assays ›
          Gene Symbol:
          TFDP2
          Gene Name:
          transcription factor Dp-2
          Gene Aliases:
          DP2
          Chromosome Location:
          Chr.3: 141944428 - 142149544 on Build GRCh38
          UniGene ID:
          Hs.379018
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n364920

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TFDP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001178138.1 NP_001171609.1 14 7005
          NM_001178139.1 NP_001171610.1 13 6967
          NM_001178140.1 NP_001171611.1 10 6463
          NM_001178141.1 NP_001171612.1 10 6396
          NM_001178142.1 NP_001171613.1 8 6244
          NM_006286.4 NP_006277.1 11 6521
          XM_005247731.2 XP_005247788.1 13 6970
          XM_011513101.2 XP_011511403.1 14 7066
          XM_011513102.2 XP_011511404.1 12 6639
          XM_011513106.2 XP_011511408.1 14 7063
          XM_011513108.2 XP_011511410.1 11 6793
          XM_011513109.2 XP_011511411.1 9 6365
          XM_011513111.2 XP_011511413.1 9 6362
          XM_017007091.1 XP_016862580.1 13 6978
          XM_017007092.1 XP_016862581.1 16 7224
          XM_017007093.1 XP_016862582.1 16 7413
          XM_017007094.1 XP_016862583.1 13 6999
          XM_017007095.1 XP_016862584.1 15 7189
          XM_017007096.1 XP_016862585.1 12 6896
          XM_017007097.1 XP_016862586.1 14 6995
          XM_017007098.1 XP_016862587.1 14 7143
          XM_017007099.1 XP_016862588.1 15 7118
          XM_017007101.1 XP_016862590.1 13 6951
          XM_017007102.1 XP_016862591.1 11 6746
          GenBank mRNA AK096450.1 - 1 51
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BF797567.1 - 1 12 Chr. 3 - 141944719 - 141947352 [-] on Build GRCh38
          GenBankmRNA BM464287.1 - 303 Chr. - - [-] on Build
          GenBankmRNA DA766812.1 - 1 51 Chr. 3 - 141946820 - 141947391 [-] on Build GRCh38
          GenBankmRNA HY114495.1 - 1 50 Chr. 3 - 141946999 - 141947390 [-] on Build GRCh38
          GenBankmRNA DA233188.1 - 2 304 Chr. 3 - 141947046 - 141948656 [-] on Build GRCh38
          GenBankmRNA DB135683.1 - 1 86 Chr. 3 - 141946975 - 141947426 [-] on Build GRCh38
          GenBankmRNA HY104526.1 - 1 175 Chr. 3 - 141947053 - 141947515 [-] on Build GRCh38
          GenBankmRNA DB155360.1 - 1 179 Chr. 3 - 141946937 - 141947506 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          negative regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription cofactor activity
          protein binding
          transcription factor binding
          protein domain specific binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0