Search
Search

CALML3-AS1| Gene Symbol: CALML3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120496.1 | 4 | 507 | |||
| NR_120497.1 | 3 | 395 | ||||
| GenBank mRNA | AK128534.1 | - | 2 | 1329 | ||
| DA220455.1 | - | 3 | 397 | |||
| DA770774.1 | - | 4 | 502 | |||