Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MELTF SIRNA Assays ›
          Gene Symbol:
          MELTF
          Gene Name:
          melanotransferrin
          Gene Aliases:
          CD228, MAP97, MFI2, MTF1, MTf
          Chromosome Location:
          Chr.3: 197001740 - 197029816 on Build GRCh38
          UniGene ID:
          Hs.184727
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n373032

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MELTF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005929.5 NP_005920.2 16 3769
          XM_006713643.3 XP_006713706.2 16 3846
          XM_011512850.2 XP_011511152.1 16 3849
          GenBank mRNA BC034933.1 - 1 743
          BC068085.1 - 1 515
          BC082761.1 - 1 515
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM749707.1 - 1 393 Chr. 3 - 197001792 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757703.1 - 1 393 Chr. 3 - 197001783 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM758731.1 - 1 393 Chr. 3 - 197001793 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757642.1 - 1 393 Chr. 3 - 197001773 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757674.1 - 1 393 Chr. 3 - 197001782 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM758279.1 - 1 393 Chr. 3 - 197001787 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM758746.1 - 1 393 Chr. 3 - 197001783 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM756635.1 - 1 393 Chr. 3 - 197001782 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757665.1 - 1 393 Chr. 3 - 197001774 - 197002326 [-] on Build GRCh38
          GenBankmRNA BP391592.1 - 1 210 Chr. 3 - 197001741 - 197002142 [-] on Build GRCh38
          GenBankmRNA BM758491.1 - 1 393 Chr. 3 - 197001772 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM750636.1 - 1 393 Chr. 3 - 197001769 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM757689.1 - 1 393 Chr. 3 - 197001773 - 197002326 [-] on Build GRCh38
          GenBankmRNA AA291458.1 - 1 113 Chr. 3 - 197001746 - 197002042 [-] on Build GRCh38
          GenBankmRNA BM743332.1 - 1 393 Chr. 3 - 197001787 - 197002326 [-] on Build GRCh38
          GenBankmRNA BM748658.1 - 1 393 Chr. 3 - 197001771 - 197002326 [-] on Build GRCh38
          GenBankmRNA BU172111.1 - 1 513 Chr. 3 - 197001742 - 197002446 [-] on Build GRCh38
          GenBankmRNA BM758998.1 - 1 392 Chr. 3 - 197001783 - 197002326 [-] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell growth
          positive regulation of plasminogen activation
          regulation of cell proliferation
          iron ion homeostasis
          positive regulation of extracellular matrix disassembly
          iron ion import
          negative regulation of substrate adhesion-dependent cell spreading
          iron ion binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-zzfgb:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline