Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TEX261 SIRNA Assays ›
          Gene Symbol:
          Tex261
          Gene Name:
          testis expressed gene 261
          Gene Aliases:
          3110001O07Rik, AA409339, AI480706, AL033351, TEG-261
          Chromosome Location:
          Chr.6: 83768881 - 83775814 on Build GRCm38
          UniGene ID:
          Mm.28371
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n403087

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tex261
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK013969.1 - 1 1000
          AK013971.1 - 1 1004
          AK170049.1 - 1 1192
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CK627063.1 - 1 298 Chr. 6 - 83769005 - 83769637 [-] on Build GRCm38
          GenBankmRNA BB681549.1 - 1 45 Chr. 6 - 83768892 - 83769384 [-] on Build GRCm38
          GenBankmRNA CR513842.1 - 1 119 Chr. 6 - 83769113 - 83769458 [-] on Build GRCm38
          GenBankmRNA AA856298.1 - 1 321 Chr. 6 - 83769236 - 83769660 [-] on Build GRCm38
          GenBankmRNA W80179.1 - 183 Chr. - - [-] on Build
          GenBankmRNA AV460057.1 - 1 443 Chr. 6 - 83769317 - 83769782 [-] on Build GRCm38
          GenBankmRNA BI872980.1 - 1 477 Chr. 6 - 83768901 - 83769817 [-] on Build GRCm38
          GenBankmRNA CJ238675.1 - 1 236 Chr. 6 - 83768884 - 83769573 [-] on Build GRCm38
          GenBankmRNA BF122781.1 - 1 219 Chr. 6 - 83768894 - 83769559 [-] on Build GRCm38
          GenBankmRNA BY763821.1 - 1 236 Chr. 6 - 83768890 - 83769573 [-] on Build GRCm38
          GenBankmRNA BI648902.1 - 1 281 Chr. 6 - 83768881 - 83769620 [-] on Build GRCm38
          GenBankmRNA BE534642.1 - 1 498 Chr. 6 - 83769252 - 83769836 [-] on Build GRCm38
          GenBankmRNA BE915641.1 - 1 124 Chr. 6 - 83768892 - 83769465 [-] on Build GRCm38
          GenBankmRNA BE687239.1 - 1 338 Chr. 6 - 83769135 - 83769677 [-] on Build GRCm38
          GenBankmRNA AA217320.1 - 1 103 Chr. 6 - 83768946 - 83769442 [-] on Build GRCm38
          GenBankmRNA BB346685.2 - 1 222 Chr. 6 - 83768890 - 83769531 [-] on Build GRCm38
          GenBankmRNA BY473746.1 - 1 22 Chr. 6 - 83768886 - 83769361 [-] on Build GRCm38
          GenBankmRNA AI159531.1 - 1 143 Chr. 6 - 83769135 - 83769478 [-] on Build GRCm38
          GenBankmRNA CB321552.1 - 1 393 Chr. 6 - 83768947 - 83769732 [-] on Build GRCm38
          GenBankmRNA BI453188.1 - 184 Chr. - - [-] on Build
          GenBankmRNA BI689378.1 - 1 98 Chr. 6 - 83768888 - 83769401 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0