Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TMEM164 SIRNA Assays ›
          Gene Symbol:
          Tmem164
          Gene Name:
          transmembrane protein 164
          Gene Aliases:
          AI316850, AI852450, AW547186, F730011B02
          Chromosome Location:
          Chr.X: 142680347 - 142843494 on Build GRCm38
          UniGene ID:
          Mm.249648
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n408906

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tmem164
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK159942.1 - 1 3577
          AK161476.1 - 1 2780
          BC059043.1 - 1 2876
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BB097151.1 - 1 248 Chr. X - 142838047 - 142838369 [+] on Build GRCm38
          GenBankmRNA BF464704.1 - 1 93 Chr. X - 142838202 - 142838367 [+] on Build GRCm38
          GenBankmRNA BY415605.1 - 1 243 Chr. X - 142838052 - 142838369 [+] on Build GRCm38
          GenBankmRNA BI694207.1 - 1 460 Chr. X - 142837832 - 142838371 [+] on Build GRCm38
          GenBankmRNA BB758691.1 - 1 452 Chr. X - 142837851 - 142838367 [+] on Build GRCm38
          GenBankmRNA CX223200.1 - 106 Chr. - - [-] on Build
          GenBankmRNA BI154518.1 - 1 407 Chr. X - 142837888 - 142838359 [+] on Build GRCm38
          GenBankmRNA BB752871.1 - 1 210 Chr. X - 142838085 - 142838364 [+] on Build GRCm38
          GenBankmRNA BB819399.1 - 1 341 Chr. X - 142837962 - 142838369 [+] on Build GRCm38
          GenBankmRNA CJ309988.1 - 1 321 Chr. X - 142837974 - 142838369 [+] on Build GRCm38
          GenBankmRNA CJ302779.1 - 1 347 Chr. X - 142837948 - 142838369 [+] on Build GRCm38
          GenBankmRNA BY667237.1 - 1 301 Chr. X - 142837994 - 142838369 [+] on Build GRCm38
          GenBankmRNA CF538220.1 - 1 99 Chr. X - 142838196 - 142838371 [+] on Build GRCm38
          GenBankmRNA BY427739.1 - 1 336 Chr. X - 142837960 - 142838369 [+] on Build GRCm38
          GenBankmRNA BB798123.1 - 1 328 Chr. X - 142837967 - 142838358 [+] on Build GRCm38
          GenBankmRNA BY636428.1 - 1 314 Chr. X - 142837982 - 142838369 [+] on Build GRCm38
          GenBankmRNA CN458912.1 - 1 638 Chr. X - 142837657 - 142838351 [+] on Build GRCm38
          GenBankmRNA BY663567.1 - 1 310 Chr. X - 142837986 - 142838369 [+] on Build GRCm38
          GenBankmRNA BY430980.1 - 1 341 Chr. X - 142837954 - 142838369 [+] on Build GRCm38
          GenBankmRNA BB791451.1 - 1 229 Chr. X - 142838092 - 142838367 [+] on Build GRCm38
          GenBankmRNA BY484358.1 - 1 288 Chr. X - 142838006 - 142838368 [+] on Build GRCm38
          GenBankmRNA CJ314123.1 - 1 314 Chr. X - 142837981 - 142838369 [+] on Build GRCm38
          GenBankmRNA CJ312834.1 - 1 507 Chr. X - 142837788 - 142838369 [+] on Build GRCm38
          GenBankmRNA BB820749.1 - 1 344 Chr. X - 142837954 - 142838374 [+] on Build GRCm38
          GenBankmRNA BY477517.1 - 1 446 Chr. X - 142837848 - 142838369 [+] on Build GRCm38
          GenBankmRNA BB801074.1 - 1 311 Chr. X - 142837985 - 142838359 [+] on Build GRCm38
          GenBankmRNA BB811787.1 - 1 347 Chr. X - 142837948 - 142838369 [+] on Build GRCm38
          GenBankmRNA BB804873.1 - 1 317 Chr. X - 142837983 - 142838358 [+] on Build GRCm38
          GenBankmRNA CJ323214.1 - 1 397 Chr. X - 142837898 - 142838369 [+] on Build GRCm38
          GenBankmRNA BY518121.1 - 1 235 Chr. X - 142838115 - 142838367 [+] on Build GRCm38
          GenBankmRNA CJ300662.1 - 1 385 Chr. X - 142837910 - 142838369 [+] on Build GRCm38
          GenBankmRNA CJ235530.1 - 1 619 Chr. X - 142837676 - 142838367 [+] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0