Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SCAF8 SIRNA Assays ›
          Gene Symbol:
          Scaf8
          Gene Name:
          SR-related CTD-associated factor 8
          Gene Aliases:
          A630086M08Rik, A930036P18Rik, AA408306, AI447644, AI448652, AU015035, Rbm16, mKIAA1116
          Chromosome Location:
          Chr.17: 3114972 - 3198859 on Build GRCm38
          UniGene ID:
          Mm.124373
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n411686

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Scaf8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK020937.1 - 296
          AK085429.1 - 573
          AK162043.1 - 904
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BB283932.1 - 1 207 Chr. 17 - 3258256 - 3258771 [+] on Build GRCm38
          GenBankmRNA BB500071.2 - 1 293 Chr. 17 - 3258155 - 3258807 [+] on Build GRCm38
          GenBankmRNA AW681705.1 - 1 124 Chr. 17 - 3258343 - 3258887 [+] on Build GRCm38
          GenBankmRNA BB661156.1 - 1 570 Chr. 17 - 3257880 - 3258500 [+] on Build GRCm38
          GenBankmRNA BQ748526.1 - 1 98 Chr. 17 - 3258351 - 3258897 [+] on Build GRCm38
          GenBankmRNA BE849143.1 - 1 85 Chr. 17 - 3258376 - 3258753 [+] on Build GRCm38
          GenBankmRNA BG076672.2 - 1 69 Chr. 17 - 3258378 - 3258769 [+] on Build GRCm38
          GenBankmRNA BE849131.1 - 1 71 Chr. 17 - 3258376 - 3258753 [+] on Build GRCm38
          GenBankmRNA AA881647.1 - 1 63 Chr. 17 - 3258384 - 3258771 [+] on Build GRCm38
          GenBankmRNA BB667204.1 - 1 148 Chr. 17 - 3258298 - 3258897 [+] on Build GRCm38
          GenBankmRNA AA409531.1 - 1 73 Chr. 17 - 3258374 - 3258723 [+] on Build GRCm38
          GenBankmRNA BB284495.1 - 1 68 Chr. 17 - 3258379 - 3258771 [+] on Build GRCm38
          GenBankmRNA AI060709.1 - 1 98 Chr. 17 - 3258350 - 3258769 [+] on Build GRCm38
          GenBankmRNA BX518132.1 - 1 97 Chr. 17 - 3258359 - 3258783 [+] on Build GRCm38
          GenBankmRNA BE689561.1 - 1 178 Chr. 17 - 3258269 - 3258736 [+] on Build GRCm38
          GenBankmRNA CB524813.1 - 1 287 Chr. 17 - 3258213 - 3258810 [+] on Build GRCm38
          GenBankmRNA BB284308.1 - 1 98 Chr. 17 - 3258380 - 3258897 [+] on Build GRCm38
          GenBankmRNA AA408307.1 - 1 73 Chr. 17 - 3258374 - 3258671 [+] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0