Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gm2087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102437.1 | 2, 3 | 175 | |||
GenBank mRNA | AK076672.1 | - | 2, 3 | 177 | ||
AV260139.2 | - | 2, 3 | 192 | |||
BB614956.1 | - | 2, 3 | 177 |