Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MAGT1 SIRNA Assays ›
          Gene Symbol:
          Magt1
          Gene Name:
          magnesium transporter 1
          Gene Aliases:
          2410001C15Rik, 2610529C04Rik, 2810482I07Rik, IAG2, IAP
          Chromosome Location:
          Chr.X: 105968084 - 106011899 on Build GRCm38
          UniGene ID:
          Mm.275943
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n431434

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 404.00
          Your Price
          Online offer: 391.65
          391.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Magt1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK013434.1 - 1 1228
          AV024949.2 - 1 852
          BC023242.1 - 1 207
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA EH094033.1 - 1 274 Chr. X - 105968204 - 105968510 [-] on Build GRCm38
          GenBankmRNA BY679921.1 - 1 221 Chr. X - 105968087 - 105968472 [-] on Build GRCm38
          GenBankmRNA CF751765.1 - 1 558 Chr. X - 105968084 - 105968809 [-] on Build GRCm38
          GenBankmRNA CJ219286.1 - 1 241 Chr. X - 105968088 - 105968498 [-] on Build GRCm38
          GenBankmRNA BI456382.1 - 1 559 Chr. X - 105968079 - 105968810 [-] on Build GRCm38
          GenBankmRNA BF661827.1 - 1 360 Chr. X - 105968182 - 105968611 [-] on Build GRCm38
          GenBankmRNA BB728184.1 - 1 277 Chr. X - 105968087 - 105968529 [-] on Build GRCm38
          GenBankmRNA BY669398.1 - 1 214 Chr. X - 105968085 - 105968465 [-] on Build GRCm38
          GenBankmRNA BY596809.1 - 1 300 Chr. X - 105968092 - 105968551 [-] on Build GRCm38
          GenBankmRNA CJ306179.1 - 1 246 Chr. X - 105968087 - 105968497 [-] on Build GRCm38
          GenBankmRNA BY409966.1 - 1 223 Chr. X - 105968087 - 105968474 [-] on Build GRCm38
          GenBankmRNA BY682023.1 - 1 238 Chr. X - 105968088 - 105968488 [-] on Build GRCm38
          GenBankmRNA BB812812.1 - 1 234 Chr. X - 105968087 - 105968485 [-] on Build GRCm38
          GenBankmRNA BY578955.1 - 1 242 Chr. X - 105968087 - 105968493 [-] on Build GRCm38
          GenBankmRNA BY667881.1 - 1 227 Chr. X - 105968087 - 105968474 [-] on Build GRCm38
          GenBankmRNA BG091474.1 - 1 359 Chr. X - 105968241 - 105968610 [-] on Build GRCm38
          GenBankmRNA CK636450.1 - 1 462 Chr. X - 105968087 - 105968713 [-] on Build GRCm38
          GenBankmRNA CJ322597.1 - 1 241 Chr. X - 105968087 - 105968492 [-] on Build GRCm38
          GenBankmRNA BB780645.1 - 1 291 Chr. X - 105968092 - 105968542 [-] on Build GRCm38
          GenBankmRNA BB768380.1 - 1 374 Chr. X - 105968087 - 105968623 [-] on Build GRCm38
          GenBankmRNA BY606319.1 - 1 206 Chr. X - 105968087 - 105968457 [-] on Build GRCm38
          GenBankmRNA AA245303.1 - 1 331 Chr. X - 105968134 - 105968582 [-] on Build GRCm38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0