Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: LINC01273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109943.1 | 2 | 251 | |||
GenBank mRNA | BM750089.1 | - | 2 | 232 | ||
BQ646539.1 | - | 2 | 251 |